Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_015506.2 | NP_056321.2 | 4 | 716 | |
XM_005270724.4 | XP_005270781.1 | 3 | 475 | ||
XM_011541204.2 | XP_011539506.1 | 4 | 625 | ||
GenBank mRNA | AK098537.1 | - | 4 | 716 | |
AL080062.1 | - | 4 | 587 | ||
BC006122.1 | - | 4 | 559 | 3535453 | |
BM980565.1 | - | 1 | 376 | ||
BU839693.1 | - | 4 | 571 |