Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_198944.1 | NP_945182.1 | 1 | 699 | |
XM_005259853.3 | XP_005259910.1 | 3 | 1203 | ||
XM_011527915.2 | XP_011526217.1 | 5 | 1624 | ||
GenBank mRNA | BC117212.1 | - | 1 | 774 | |
X89676.1 | - | 1 | 325 |