Gene Transcripts

Gene Symbol
RHOD
Entrez Gene ID
29984
Gene Name
ras homolog family member D
Gene Aliases
ARHD, RHOHP1, RHOM, Rho
Chromosome Location
Chr. 11: 67056818 - 67072017 on Build GRCh38
UniGene ID
Hs.15114
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014578.3 NP_055393.1 3, 4 424
GenBank mRNA AF498973.1 - 3, 4 339
BC001338.1 - 3, 4 392 3461774
BM761362.1 - 3, 4 424

View Full Product Details