Gene Transcripts

Gene Symbol
KCP
Entrez Gene ID
375616
Gene Name
kielin/chordin-like protein
Gene Aliases
CRIM2, KCP1, NET67
Chromosome Location
Chr. 7: 128870645 - 128910719 on Build GRCh38
UniGene ID
Hs.371746
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135914.1 NP_001129386.1 15, 16 1674
NM_199349.2 NP_955381.2 17, 18 1848
XM_017012184.1 XP_016867673.1 17, 18 1848
XM_017012185.1 XP_016867674.1 17, 18 1848
XM_017012186.1 XP_016867675.1 17, 18 1848
XM_017012187.1 XP_016867676.1 17, 18 1848
XM_017012188.1 XP_016867677.1 17, 18 1848
XM_017012189.1 XP_016867678.1 17, 18 1848
XM_017012190.1 XP_016867679.1 17, 18 1848
XM_017012191.1 XP_016867680.1 10, 11 1067
XM_017012192.1 XP_016867681.1 10, 11 1078
XM_017012193.1 XP_016867682.1 17, 18 1848
XM_017012194.1 XP_016867683.1 17, 18 1848
GenBank mRNA AK095464.1 - 10, 11 1322
AK122706.1 - 17, 18 1848

View Full Product Details