Gene Transcripts
- Gene Symbol
- ARSB
- Entrez Gene ID
- 411
- Gene Name
- arylsulfatase B
- Gene Aliases
- ASB, G4S, MPS6
- Chromosome Location
- Chr. 5: 78777209 - 78986534 on Build GRCh38
- UniGene ID
- Hs.149103
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000046.3 |
NP_000037.2
|
1, 2 |
1599 |
|
NM_198709.2 |
NP_942002.1
|
2, 3 |
668 |
|
XM_011543390.1 |
XP_011541692.1
|
2, 3 |
812 |
|
XM_011543391.2 |
XP_011541693.1
|
1, 2 |
375 |
|
XM_011543392.2 |
XP_011541694.1
|
1, 2 |
375 |
|
XM_011543393.1 |
XP_011541695.1
|
1, 2 |
375 |
|
XM_017009471.1 |
XP_016864960.1
|
1, 2 |
375 |
|
GenBank mRNA |
AK290865.1 |
- |
1, 2 |
375 |
|
BC029051.1 |
- |
2, 3 |
668 |
5186657 |
J05225.1 |
- |
1, 2 |
319 |
|
M32373.1 |
- |
1, 2 |
872 |
|
View Full Product Details