Gene Transcripts

Gene Symbol
CERCAM
Entrez Gene ID
51148
Gene Name
cerebral endothelial cell adhesion molecule
Gene Aliases
CEECAM1, GLT25D3
Chromosome Location
Chr. 9: 128405993 - 128437351 on Build GRCh38
UniGene ID
Hs.495230
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286760.1 NP_001273689.1 10 1239
NM_016174.4 NP_057258.3 10 1715
XM_005252035.1 XP_005252092.1 12 2625
XM_011518762.1 XP_011517064.1 13 2782
XM_011518763.2 XP_011517065.1 10 2333
XM_017014794.1 XP_016870283.1 14 2524
GenBank mRNA AB040935.1 - 11 4228
AF177203.1 - 9 1083
AF218016.1 - 5 465
AK074519.1 - 9 1151
AK074523.1 - 9 1125
AK297929.1 - 9 1211
AK307753.1 - 7 992
AK314294.1 - 10 1286
BC011811.2 - 3 358
BC098432.1 - 10 1313 6672104
BC108698.1 - 10 1326
BC119698.1 - 10 1188 40116832
BC119699.1 - 10 1188 40116834

View Full Product Details