Gene Transcripts

Gene Symbol
ACP6
Entrez Gene ID
51205
Gene Name
acid phosphatase 6, lysophosphatidic
Gene Aliases
ACPL1, LPAP, PACPL1
Chromosome Location
Chr. 1: 147629658 - 147670543 on Build GRCh38
UniGene ID
Hs.562154
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001323625.1 NP_001310554.1 3 879
NM_016361.4 NP_057445.4 3 848
NR_136633.1 3 879
NR_136634.1 3 1199
NR_136635.1 3 879
NR_136636.1 3 1199
XM_011509601.2 XP_011507903.1 3 804
XM_017001422.1 XP_016856911.1 3 804
GenBank mRNA AB030039.2 - 3 652
AB031478.1 - 3 759
AB209248.1 - 3 621
AK000657.1 - 3 585
AK308593.1 - 2 486
AL520992.3 - 3 847
AY358552.1 - 3 556
BC009965.1 - 3 828 4299943
BC034686.1 - 3 832 3463770
CR457101.1 - 3 363
KJ534759.1 - 3 384
KJ535036.1 - 2 256
KJ535044.1 - 3 384
KJ535055.1 - 3 384

View Full Product Details