Gene Transcripts
- Gene Symbol
- NUB1
- Entrez Gene ID
- 51667
- Gene Name
- negative regulator of ubiquitin like proteins 1
- Gene Aliases
- BS4, NUB1L, NYREN18
- Chromosome Location
- Chr. 7: 151341699 - 151378461 on Build GRCh38
- UniGene ID
- Hs.647082
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001243351.1 |
NP_001230280.1
|
11, 12 |
1337 |
|
NM_016118.4 |
NP_057202.3
|
11, 12 |
1337 |
|
XM_005250011.2 |
XP_005250068.1
|
11, 12 |
1402 |
|
XM_017012304.1 |
XP_016867793.1
|
11, 12 |
1396 |
|
XM_017012305.1 |
XP_016867794.1
|
11, 12 |
1396 |
|
XM_017012306.1 |
XP_016867795.1
|
11, 12 |
1418 |
|
XM_017012310.1 |
XP_016867799.1
|
12, 13 |
1499 |
|
XM_017012311.1 |
XP_016867800.1
|
12, 13 |
1403 |
|
XM_017012312.1 |
XP_016867801.1
|
12, 13 |
1372 |
|
XM_017012313.1 |
XP_016867802.1
|
12, 13 |
1499 |
|
XM_017012314.1 |
XP_016867803.1
|
12, 13 |
1375 |
|
GenBank mRNA |
AF155099.1 |
- |
11, 12 |
1269 |
|
AF300717.1 |
- |
11, 12 |
1326 |
|
AF459743.1 |
- |
11, 12 |
1285 |
|
AY129295.1 |
- |
11, 12 |
1285 |
|
BC046354.2 |
- |
10, 11 |
1253 |
6070128 |
View Full Product Details