Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_017785.4 | NP_060255.3 | 3 | 585 | |
XM_011534585.1 | XP_011532887.1 | 3 | 531 | ||
XM_011534586.2 | XP_011532888.1 | 3 | 467 | ||
XM_011534587.2 | XP_011532889.1 | 4 | 732 | ||
XM_011534588.2 | XP_011532890.1 | 4 | 670 | ||
XM_011534589.2 | XP_011532891.1 | 3 | 480 | ||
XM_011534590.2 | XP_011532892.1 | 2 | 221 | ||
XM_017009601.1 | XP_016865090.1 | 3 | 416 | ||
XM_017009602.1 | XP_016865091.1 | 2 | 221 | ||
GenBank mRNA | AF269167.1 | - | 3 | 476 | |
AK000371.1 | - | 2 | 311 | ||
AK304623.1 | - | 3, 4 | 483 | ||
AK308835.1 | - | 3 | 464 | ||
AK310442.1 | - | 3 | 384 | ||
AY517557.1 | - | 3 | 416 | ||
BC012568.1 | - | 3 | 451 | 4102558 |