Gene Transcripts

Gene Symbol
AGK
Entrez Gene ID
55750
Gene Name
acylglycerol kinase
Gene Aliases
CATC5, CTRCT38, MTDPS10, MULK
Chromosome Location
Chr. 7: 141551278 - 141655244 on Build GRCh38
UniGene ID
Hs.743318
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018238.3 NP_060708.1 14 1201
XM_005250023.3 XP_005250080.1 14 1097
XM_011516397.2 XP_011514699.1 14 1667
GenBank mRNA AJ278150.1 - 13 1057
AK001704.1 - 14 1069
AK222769.1 - 14 1063
AK299131.1 - 13 909
AK304502.1 - 15 1088
BC022777.1 - 13 1055 3913448

View Full Product Details