Gene Transcripts

Gene Symbol
SDHB
Entrez Gene ID
6390
Gene Name
succinate dehydrogenase complex iron sulfur subunit B
Gene Aliases
CWS2, IP, PGL4, SDH, SDH1, SDH2, SDHIP
Chromosome Location
Chr. 1: 17018722 - 17054170 on Build GRCh38
UniGene ID
Hs.465924
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003000.2 NP_002991.2 8 980
GenBank mRNA AK312056.1 - 8 842
BC007840.2 - 8 980 4124737
BU554163.1 - 6 570
D10245.1 - 7 775
M32246.1 - 7 775
U17248.1 - 8 963

View Full Product Details