Gene Transcripts
- Gene Symbol
- CLSPN
- Entrez Gene ID
- 63967
- Gene Name
- claspin
- Gene Aliases
- Chromosome Location
- Chr. 1: 35720465 - 35769985 on Build GRCh38
- UniGene ID
- Hs.175613
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001190481.1 |
NP_001177410.1
|
10, 11 |
2097 |
|
NM_022111.3 |
NP_071394.2
|
11, 12 |
2289 |
|
XM_011541938.2 |
XP_011540240.1
|
11, 12 |
2312 |
|
XM_011541939.2 |
XP_011540241.1
|
11, 12 |
2306 |
|
XM_011541940.2 |
XP_011540242.1
|
11, 12 |
2306 |
|
XM_011541941.2 |
XP_011540243.1
|
10, 11 |
2144 |
|
GenBank mRNA |
BC113116.1 |
- |
11, 12 |
2282 |
|
BC115025.1 |
- |
10, 11 |
2090 |
7961363 |
BC115026.1 |
- |
11, 12 |
2282 |
7961366 |
BC137279.1 |
- |
11, 12 |
2268 |
9021276 |
BC140789.1 |
- |
11, 12 |
2268 |
9021657 |
View Full Product Details