Gene Transcripts
- Gene Symbol
- BRD9
- Entrez Gene ID
- 65980
- Gene Name
- bromodomain containing 9
- Gene Aliases
- LAVS3040, PRO9856
- Chromosome Location
- Chr. 5: 863735 - 892824 on Build GRCh38
- UniGene ID
- Hs.449278
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001009877.2 |
NP_001009877.2
|
7, 8 |
818 |
|
NM_001317951.1 |
NP_001304880.1
|
7, 8 |
1170 |
|
NM_023924.4 |
NP_076413.3
|
7, 8 |
991 |
|
NR_134296.1 |
|
7, 8 |
1327 |
|
XM_006714494.3 |
XP_006714557.1
|
7, 8 |
1030 |
|
GenBank mRNA |
AK023503.1 |
- |
7, 8 |
1148 |
|
AK024392.1 |
- |
4, 5 |
826 |
|
AK026830.1 |
- |
2, 3 |
122 |
|
AK297573.1 |
- |
7, 8 |
805 |
|
AK299157.1 |
- |
7, 8 |
991 |
|
AK301987.1 |
- |
7, 8 |
942 |
|
BC007217.1 |
- |
7, 8 |
1007 |
|
BC041590.1 |
- |
5, 6 |
677 |
5428011 |
BX648703.1 |
- |
7, 8 |
1329 |
|
DQ248311.1 |
- |
3, 4 |
316 |
|
View Full Product Details