Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_003088.3 | NP_003079.1 | 5 | 1475 | |
GenBank mRNA | AK095254.1 | - | 6 | 1409 | |
AK095830.1 | - | 4 | 896 | ||
AK125295.1 | - | 1 | 672 | ||
AK316607.1 | - | 5 | 1470 | ||
BC000521.2 | - | 5 | 1366 | 2961047 | |
BC006304.2 | - | 5 | 1457 | 4100996 | |
BC007539.1 | - | 5 | 1366 | ||
BC007643.1 | - | 5 | 1099 | ||
BC007948.2 | - | 5 | 1454 | 4299524 | |
BC007988.2 | - | 5 | 1366 | 2961047 | |
BT006636.1 | - | 5 | 1343 | ||
U03057.1 | - | 5 | 1454 | ||
U09873.1 | - | 5 | 1388 |