Gene Transcripts

Gene Symbol
SNRNP70
Entrez Gene ID
6625
Gene Name
small nuclear ribonucleoprotein U1 subunit 70
Gene Aliases
RNPU1Z, RPU1, SNRP70, Snp1, U1-70K, U170K, U1AP, U1RNP
Chromosome Location
Chr. 19: 49085140 - 49108613 on Build GRCh38
UniGene ID
Hs.467097
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001301069.1 NP_001287998.1 10 1761
NM_003089.5 NP_003080.2 10 1788
XM_005259178.1 XP_005259235.1 4 821
XM_011527240.2 XP_011525542.2 5 1007
XM_011527241.2 XP_011525543.1 4 974
GenBank mRNA AK027127.1 - 3 1314
AK056808.1 - 3 2878
AK096783.1 - 11 2951
AK293120.1 - 10 1507
AL117399.1 - 2 2780
AL117507.1 - 3 1820
AL832819.1 - 3 3029
BC000342.2 - 2 682
BC001315.1 - 10 1461 3454479
BM969999.1 - 1 218
M22636.1 - 10 1452
X06811.1 - 1 196
X06815.1 - 11 1477
X06816.1 - 1 323
X07401.1 - 4 827
X07402.1 - 1 206
X84841.1 - 9 1281

View Full Product Details