Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005633.3 | NP_005624.2 | 13 | 2131 | |
XM_005264515.4 | XP_005264572.1 | 13 | 2360 | ||
XM_011533062.2 | XP_011531364.1 | 13 | 2299 | ||
XM_011533064.2 | XP_011531366.1 | 13 | 2039 | ||
XM_011533066.2 | XP_011531368.1 | 6 | 1040 | ||
GenBank mRNA | AB209140.1 | - | 6 | 1102 | |
AK290228.1 | - | 14 | 2177 | ||
L13857.1 | - | 13 | 2090 |