Gene Transcripts

Gene Symbol
CENPM
Entrez Gene ID
79019
Gene Name
centromere protein M
Gene Aliases
C22orf18, CENP-M, PANE1
Chromosome Location
Chr. 22: 41927747 - 41947164 on Build GRCh38
UniGene ID
Hs.208912
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001002876.2 NP_001002876.1 3 290
NM_001304370.1 NP_001291299.1 2 403
NM_001304371.1 NP_001291300.1 3 290
NM_001304372.1 NP_001291301.1 2 403
NM_001304373.1 NP_001291302.1 2 403
NM_024053.4 NP_076958.1 3 290
XM_011530368.2 XP_011528670.1 3 277
GenBank mRNA AK123479.1 - 3 279
AL160131.1 - 3 238
BC000705.2 - 3 237 3349507
BC007495.2 - 3 250 2966541
BE797487.1 - 3 244
BM045658.1 - 2 378
BQ652721.1 - 2 240
BQ922403.1 - 3 223
BU556693.1 - 3 290
BX332140.2 - 2 341
BX354019.2 - 2 337
BX417558.2 - 2 342
CA414668.1 - 2 424
CR456399.1 - 3 263
CR457392.1 - 3 202
HY095078.1 - 2 404

View Full Product Details