Gene Transcripts
- Gene Symbol
- TMEM243
- Entrez Gene ID
- 79161
- Gene Name
- transmembrane protein 243
- Gene Aliases
- C7orf23, MM-TRAG, MMTRAG
- Chromosome Location
- Chr. 7: 87195593 - 87220603 on Build GRCh38
- UniGene ID
- Hs.619593
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
XM_005250586.3 |
XP_005250643.1
|
3, 4 |
551 |
|
XM_005250587.4 |
XP_005250644.1
|
3, 4 |
685 |
|
XM_005250588.3 |
XP_005250645.1
|
3, 4 |
383 |
|
XM_005250589.3 |
XP_005250646.1
|
3, 4 |
551 |
|
XM_017012616.1 |
XP_016868105.1
|
2, 3 |
972 |
|
XM_017012617.1 |
XP_016868106.1
|
3, 4 |
674 |
|
XM_017012618.1 |
XP_016868107.1
|
3, 4 |
418 |
|
GenBank mRNA |
AK313794.1 |
- |
2, 3 |
353 |
|
AL711214.1 |
- |
3, 4 |
389 |
|
BC002837.2 |
- |
2, 3 |
335 |
3634983 |
BP236768.1 |
- |
3, 4 |
442 |
|
BT006973.1 |
- |
2, 3 |
123 |
|
CB993361.1 |
- |
3, 4 |
592 |
|
DB241717.1 |
- |
2, 3 |
222 |
|
View Full Product Details