Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005677.3 | NP_005668.2 | 2 | 314 | |
NM_080538.2 | NP_536799.1 | 2 | 443 | ||
NM_080539.3 | NP_536800.2 | 2 | 314 | ||
GenBank mRNA | AF057036.1 | - | 2 | 314 | |
AJ225895.1 | - | 2 | 307 | ||
AK128401.1 | - | 2 | 443 | ||
AK289688.1 | - | 2 | 333 | ||
AY150334.1 | - | 2 | 291 | ||
AY150335.1 | - | 2 | 291 | ||
AY150336.1 | - | 2 | 291 | ||
AY150337.1 | - | 2 | 291 | ||
AY150338.1 | - | 2 | 291 | ||
AY150339.1 | - | 2 | 291 | ||
BC074828.2 | - | 2 | 218 | 30915542 | |
BC074829.2 | - | 2 | 218 | 30915325 | |
BX500613.1 | - | 2 | 301 |