Gene Transcripts

Gene Symbol
YPEL3
Entrez Gene ID
83719
Gene Name
yippee like 3
Gene Aliases
Chromosome Location
Chr. 16: 30092314 - 30096216 on Build GRCh38
UniGene ID
Hs.513491
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145524.1 NP_001138996.1 5 513
NM_031477.4 NP_113665.3 4 1159
GenBank mRNA AB098737.1 - 5 513
AF305622.1 - 5 516
BC005009.1 - 5 469 3639517
BC050664.1 - 4 1158 5748830

View Full Product Details