Gene Transcripts

Gene Symbol
PLEKHN1
Entrez Gene ID
84069
Gene Name
pleckstrin homology domain containing N1
Gene Aliases
Chromosome Location
Chr. 1: 965820 - 975108 on Build GRCh38
UniGene ID
Hs.528691
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001160184.1 NP_001153656.1 2 191
NM_032129.2 NP_115505.2 2 191
XM_006710944.3 XP_006711007.2 2 868
XM_011542248.2 XP_011540550.2 2 868
XM_017002474.1 XP_016857963.1 2 868
XM_017002475.1 XP_016857964.1 2 868
XM_017002476.1 XP_016857965.1 2 868
XM_017002477.1 XP_016857966.1 2 868
XM_017002478.1 XP_016857967.1 2 868
XM_017002479.1 XP_016857968.1 2 868
GenBank mRNA AL136730.1 - 2 191
BC101386.2 - 2 186 40026400
BC101387.2 - 2 186 40026404

View Full Product Details