Gene Transcripts

Gene Symbol
PCGF1
Entrez Gene ID
84759
Gene Name
polycomb group ring finger 1
Gene Aliases
2010002K04Rik, NSPC1, RNF3A-2, RNF68
Chromosome Location
Chr. 2: 74505043 - 74508580 on Build GRCh38
UniGene ID
Hs.316750
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032673.2 NP_116062.2 2 160
GenBank mRNA AF087884.1 - 1 266
AK125742.1 - 2 1046
BC004952.1 - 2 125 3621400
BU602498.1 - 2 163

View Full Product Details