Gene Transcripts

Gene Symbol
RHOBTB1
Entrez Gene ID
9886
Gene Name
Rho related BTB domain containing 1
Gene Aliases
Chromosome Location
Chr. 10: 60869438 - 61001486 on Build GRCh38
UniGene ID
Hs.737374
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001242359.1 NP_001229288.1 9 2114
NM_014836.4 NP_055651.1 8 2042
NR_024554.2 9 1916
NR_024555.1 9 1880
XM_006718084.1 XP_006718147.1 8 1850
XM_006718085.1 XP_006718148.1 8 1877
XM_011540424.1 XP_011538726.1 9 1908
XM_011540425.1 XP_011538727.1 9 1908
XM_011540426.1 XP_011538728.1 9 1908
XM_017017000.1 XP_016872489.1 8 2097
XM_017017001.1 XP_016872490.1 8 2093
XM_017017002.1 XP_016872491.1 8 2093
XM_017017003.1 XP_016872492.1 10 1983
XM_017017004.1 XP_016872493.1 10 1985
XM_017017005.1 XP_016872494.1 9 1890
XM_017017006.1 XP_016872495.1 9 1939
XM_017017007.1 XP_016872496.1 7 1989
XM_017017008.1 XP_016872497.1 9 2150
GenBank mRNA AB018283.3 - 8 1933
AK075129.1 - 9 1880
AK307630.1 - 7 1612
AK309440.1 - 1 84
BC032848.1 - 9 2091 4830471
BC041791.1 - 8 1833 5297756

View Full Product Details