Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 85957907-85958409 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PICALM
Entrez Gene ID
8301
Gene Name
phosphatidylinositol binding clathrin assembly protein
Gene Aliases
CALM, CLTH, LAP
UniGene
Hs.163893
Gene Chromosome Location
Chr.11: 85957171-86069881 on Build GRCh38
Transcripts
NM_001008660.2 NM_001206946.1 NM_001206947.1 NM_007166.3 XM_005274322.2 XM_005274323.2 XM_005274324.2 XM_005274325.2 XM_005274326.2 XM_005274327.2 XM_005274328.2 XM_005274329.3 XM_005274330.2 XM_005274331.2 XM_005274332.2 XM_005274333.2 XM_005274334.2 XM_005274335.2 XM_005274336.2 XM_005274337.2 XM_005274340.2 XM_006718699.2 XM_006718700.2 XM_006718701.3 XM_011545293.1 XM_017018381.1 XM_017018382.1 XM_017018383.1 XM_017018384.1 XM_017018385.1 XM_017018386.1 XM_017018387.1 XM_017018388.1

SNP Detail

SNP ID Chromosome Location
rs906488449 Chr.11: 85958069-85958069 on Build GRCh38
rs765260263 Chr.11: 85958307-85958307 on Build GRCh38
rs893460309 Chr.11: 85958177-85958177 on Build GRCh38
rs924761117 Chr.11: 85958354-85958354 on Build GRCh38
rs562451968 Chr.11: 85957990-85957990 on Build GRCh38
rs1041851209 Chr.11: 85958083-85958083 on Build GRCh38
rs930104594 Chr.11: 85958023-85958023 on Build GRCh38
rs1017945724 Chr.11: 85958303-85958303 on Build GRCh38
rs991476373 Chr.11: 85958309-85958309 on Build GRCh38
rs1025989117 Chr.11: 85957956-85957963 on Build GRCh38
rs965386274 Chr.11: 85958306-85958306 on Build GRCh38
rs867882955 Chr.11: 85958048-85958048 on Build GRCh38
rs549795962 Chr.11: 85958265-85958265 on Build GRCh38
rs138705131 Chr.11: 85958154-85958154 on Build GRCh38
rs549111531 Chr.11: 85958174-85958174 on Build GRCh38
rs1056951670 Chr.11: 85958143-85958143 on Build GRCh38
rs1006536748 Chr.11: 85958192-85958192 on Build GRCh38
rs766554201 Chr.11: 85958215-85958215 on Build GRCh38
rs560065389 Chr.11: 85958160-85958160 on Build GRCh38
rs971503670 Chr.11: 85958337-85958337 on Build GRCh38
rs1025924702 Chr.11: 85958341-85958341 on Build GRCh38
rs77343535 Chr.11: 85958261-85958261 on Build GRCh38
rs544153970 Chr.11: 85957985-85957985 on Build GRCh38
rs937456073 Chr.11: 85958033-85958033 on Build GRCh38
rs917325944 Chr.11: 85958334-85958334 on Build GRCh38
rs976585583 Chr.11: 85957932-85957932 on Build GRCh38
rs556314944 Chr.11: 85958026-85958027 on Build GRCh38
rs951595425 Chr.11: 85958355-85958355 on Build GRCh38
rs909995918 Chr.11: 85958046-85958046 on Build GRCh38
rs1045112123 Chr.11: 85958060-85958060 on Build GRCh38
rs950276362 Chr.11: 85958081-85958081 on Build GRCh38
rs918771868 Chr.11: 85958019-85958019 on Build GRCh38
rs1039874336 Chr.11: 85958198-85958198 on Build GRCh38
rs185162245 Chr.11: 85958142-85958142 on Build GRCh38
rs984322164 Chr.11: 85958359-85958359 on Build GRCh38
rs910780159 Chr.11: 85958376-85958376 on Build GRCh38
rs757491737 Chr.11: 85957962-85957973 on Build GRCh38
rs1021402287 Chr.11: 85958204-85958204 on Build GRCh38
rs533066609 Chr.11: 85958145-85958145 on Build GRCh38
rs951259611 Chr.11: 85958016-85958016 on Build GRCh38
rs1025471852 Chr.11: 85958264-85958264 on Build GRCh38
rs991964049 Chr.11: 85958045-85958045 on Build GRCh38
rs984269921 Chr.11: 85958026-85958026 on Build GRCh38
rs900887694 Chr.11: 85958249-85958249 on Build GRCh38
rs527414572 Chr.11: 85958169-85958169 on Build GRCh38
rs773609423 Chr.11: 85958137-85958137 on Build GRCh38
rs765254297 Chr.11: 85958022-85958022 on Build GRCh38
rs567326279 Chr.11: 85958178-85958178 on Build GRCh38
rs1053478155 Chr.11: 85958164-85958164 on Build GRCh38
rs762345946 Chr.11: 85958000-85958000 on Build GRCh38
rs998465990 Chr.11: 85958318-85958318 on Build GRCh38
rs754106264 Chr.11: 85958235-85958235 on Build GRCh38
rs767919632 Chr.11: 85958131-85958131 on Build GRCh38

View Full Product Details