Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 12188463-12188955 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TNFRSF1B
Entrez Gene ID
7133
Gene Name
TNF receptor superfamily member 1B
Gene Aliases
CD120b, TBPII, TNF-R-II, TNF-R75, TNFBR, TNFR1B, TNFR2, TNFR80, p75, p75TNFR
UniGene
Hs.256278
Gene Chromosome Location
Chr.1: 12166948-12209222 on Build GRCh38
Transcripts
NM_001066.2 XM_011542060.2 XM_011542063.2 XM_017002211.1 XM_017002212.1 XM_017002213.1 XM_017002214.1 XM_017002215.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
894797 c.109G>A p.G37R Chr.1: 12188826-12188826 on Build GRCh38
4408863 c.178+8C>A p.? Chr.1: 12188903-12188903 on Build GRCh38
340524 c.134A>T p.Y45F Chr.1: 12188851-12188851 on Build GRCh38
7214600 c.150T>C p.A50A Chr.1: 12188867-12188867 on Build GRCh38
6057764 c.147A>T p.T49T Chr.1: 12188864-12188864 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs962913643 Chr.1: 12188848-12188848 on Build GRCh38
rs138534863 Chr.1: 12188765-12188765 on Build GRCh38
rs139221787 Chr.1: 12188890-12188890 on Build GRCh38
rs941581673 Chr.1: 12188655-12188655 on Build GRCh38
rs777952063 Chr.1: 12188757-12188757 on Build GRCh38
rs775933189 Chr.1: 12188898-12188898 on Build GRCh38
rs1006272458 Chr.1: 12188788-12188788 on Build GRCh38
rs1000128688 Chr.1: 12188559-12188559 on Build GRCh38
rs75930962 Chr.1: 12188935-12188935 on Build GRCh38
rs933240444 Chr.1: 12188724-12188724 on Build GRCh38
rs781514091 Chr.1: 12188831-12188831 on Build GRCh38
rs780866089 Chr.1: 12188759-12188759 on Build GRCh38
rs749226357 Chr.1: 12188872-12188872 on Build GRCh38
rs540459773 Chr.1: 12188814-12188814 on Build GRCh38
rs770594342 Chr.1: 12188560-12188560 on Build GRCh38
rs762824049 Chr.1: 12188772-12188772 on Build GRCh38
rs768521380 Chr.1: 12188879-12188879 on Build GRCh38
rs758938065 Chr.1: 12188823-12188823 on Build GRCh38
rs760020939 Chr.1: 12188818-12188818 on Build GRCh38
rs747230676 Chr.1: 12188758-12188758 on Build GRCh38
rs925399356 Chr.1: 12188925-12188925 on Build GRCh38
rs374141767 Chr.1: 12188919-12188919 on Build GRCh38
rs17883721 Chr.1: 12188536-12188536 on Build GRCh38
rs377349339 Chr.1: 12188825-12188825 on Build GRCh38
rs1029360001 Chr.1: 12188592-12188592 on Build GRCh38
rs775453272 Chr.1: 12188768-12188768 on Build GRCh38
rs5746003 Chr.1: 12188682-12188682 on Build GRCh38
rs913889137 Chr.1: 12188656-12188656 on Build GRCh38
rs747763895 Chr.1: 12188909-12188910 on Build GRCh38
rs746051386 Chr.1: 12188838-12188838 on Build GRCh38
rs371638252 Chr.1: 12188914-12188914 on Build GRCh38
rs763571307 Chr.1: 12188907-12188907 on Build GRCh38
rs1033895525 Chr.1: 12188586-12188586 on Build GRCh38
rs761386236 Chr.1: 12188813-12188813 on Build GRCh38
rs997632418 Chr.1: 12188498-12188498 on Build GRCh38
rs962863464 Chr.1: 12188563-12188563 on Build GRCh38
rs140531887 Chr.1: 12188893-12188893 on Build GRCh38
rs376609863 Chr.1: 12188770-12188770 on Build GRCh38
rs945439 Chr.1: 12188885-12188885 on Build GRCh38
rs555785305 Chr.1: 12188739-12188739 on Build GRCh38
rs142907823 Chr.1: 12188800-12188800 on Build GRCh38
rs373387104 Chr.1: 12188819-12188819 on Build GRCh38
rs1014090497 Chr.1: 12188490-12188490 on Build GRCh38
rs191270958 Chr.1: 12188579-12188579 on Build GRCh38
rs983335857 Chr.1: 12188626-12188626 on Build GRCh38
rs757274903 Chr.1: 12188826-12188826 on Build GRCh38
rs558237743 Chr.1: 12188798-12188798 on Build GRCh38
rs979557374 Chr.1: 12188677-12188677 on Build GRCh38
rs937993377 Chr.1: 12188661-12188661 on Build GRCh38
rs750041133 Chr.1: 12188815-12188815 on Build GRCh38
rs590943 Chr.1: 12188628-12188628 on Build GRCh38
rs149960043 Chr.1: 12188891-12188891 on Build GRCh38
rs768438719 Chr.1: 12188909-12188909 on Build GRCh38
rs745737460 Chr.1: 12188760-12188760 on Build GRCh38
rs1000479454 Chr.1: 12188753-12188753 on Build GRCh38
rs17882763 Chr.1: 12188910-12188910 on Build GRCh38
rs538265107 Chr.1: 12188792-12188792 on Build GRCh38
rs945438 Chr.1: 12188908-12188908 on Build GRCh38
rs760356408 Chr.1: 12188894-12188894 on Build GRCh38
rs779781521 Chr.1: 12188849-12188849 on Build GRCh38
rs768429181 Chr.1: 12188750-12188750 on Build GRCh38
rs41278642 Chr.1: 12188660-12188660 on Build GRCh38
rs5746002 Chr.1: 12188501-12188501 on Build GRCh38
rs1005951671 Chr.1: 12188547-12188547 on Build GRCh38
rs535771252 Chr.1: 12188683-12188683 on Build GRCh38
rs1041911669 Chr.1: 12188690-12188690 on Build GRCh38
rs778009210 Chr.1: 12188824-12188824 on Build GRCh38
rs112131423 Chr.1: 12188908-12188908 on Build GRCh38
rs778420035 Chr.1: 12188664-12188664 on Build GRCh38
rs960984108 Chr.1: 12188598-12188598 on Build GRCh38
rs560783734 Chr.1: 12188839-12188839 on Build GRCh38
rs183755458 Chr.1: 12188806-12188806 on Build GRCh38
rs868173562 Chr.1: 12188722-12188722 on Build GRCh38
rs1038884778 Chr.1: 12188687-12188687 on Build GRCh38
rs371927176 Chr.1: 12188920-12188920 on Build GRCh38

View Full Product Details