Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 31260823-31261362 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B3GLCT
Entrez Gene ID
145173
Gene Name
beta 3-glucosyltransferase
Gene Aliases
B3GALTL, B3GTL, B3Glc-T, Gal-T, beta3Glc-T
UniGene
Hs.13205
Gene Chromosome Location
Chr.13: 31199975-31332276 on Build GRCh38
Transcripts
NM_194318.3 XM_006719768.3 XM_011534936.1 XM_011534938.2 XM_017020395.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6445867 c.510C>A p.H170Q Chr.13: 31260996-31260996 on Build GRCh38
3885227 c.524C>T p.S175F Chr.13: 31261010-31261010 on Build GRCh38
5842395 c.584C>T p.P195L Chr.13: 31261070-31261070 on Build GRCh38
127410 c.525C>T p.S175S Chr.13: 31261011-31261011 on Build GRCh38
946676 c.528G>T p.E176D Chr.13: 31261014-31261014 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781296239 Chr.13: 31261000-31261000 on Build GRCh38
rs775916141 Chr.13: 31261067-31261067 on Build GRCh38
rs936153230 Chr.13: 31261050-31261050 on Build GRCh38
rs1018372305 Chr.13: 31261244-31261244 on Build GRCh38
rs545753153 Chr.13: 31261064-31261064 on Build GRCh38
rs73172886 Chr.13: 31260874-31260874 on Build GRCh38
rs770733008 Chr.13: 31261036-31261036 on Build GRCh38
rs542243712 Chr.13: 31261163-31261163 on Build GRCh38
rs1040692535 Chr.13: 31260899-31260899 on Build GRCh38
rs771056423 Chr.13: 31260921-31260921 on Build GRCh38
rs749261155 Chr.13: 31260926-31260926 on Build GRCh38
rs980711790 Chr.13: 31261038-31261038 on Build GRCh38
rs750588447 Chr.13: 31261114-31261114 on Build GRCh38
rs116639305 Chr.13: 31260990-31260990 on Build GRCh38
rs114885586 Chr.13: 31260988-31260988 on Build GRCh38
rs777525848 Chr.13: 31261127-31261127 on Build GRCh38
rs760566733 Chr.13: 31260888-31260888 on Build GRCh38
rs936498885 Chr.13: 31261058-31261058 on Build GRCh38
rs4943289 Chr.13: 31261238-31261238 on Build GRCh38
rs115265063 Chr.13: 31261021-31261021 on Build GRCh38
rs766030579 Chr.13: 31260957-31260957 on Build GRCh38
rs1037805453 Chr.13: 31261253-31261253 on Build GRCh38
rs972487783 Chr.13: 31261001-31261001 on Build GRCh38
rs575326980 Chr.13: 31261125-31261125 on Build GRCh38
rs151026975 Chr.13: 31261011-31261011 on Build GRCh38
rs747193248 Chr.13: 31260916-31260916 on Build GRCh38
rs751928458 Chr.13: 31261099-31261099 on Build GRCh38
rs148471818 Chr.13: 31260964-31260964 on Build GRCh38
rs763510934 Chr.13: 31261075-31261075 on Build GRCh38
rs560883483 Chr.13: 31261231-31261231 on Build GRCh38
rs760374280 Chr.13: 31260951-31260951 on Build GRCh38
rs558681668 Chr.13: 31260905-31260905 on Build GRCh38
rs1053526406 Chr.13: 31260855-31260855 on Build GRCh38
rs772799456 Chr.13: 31260949-31260949 on Build GRCh38
rs556735977 Chr.13: 31261217-31261218 on Build GRCh38
rs756907968 Chr.13: 31261023-31261023 on Build GRCh38
rs781675680 Chr.13: 31261321-31261321 on Build GRCh38
rs776380103 Chr.13: 31261052-31261052 on Build GRCh38
rs926478019 Chr.13: 31261045-31261045 on Build GRCh38
rs1053508652 Chr.13: 31261066-31261066 on Build GRCh38
rs746710147 Chr.13: 31261034-31261034 on Build GRCh38
rs141304626 Chr.13: 31261254-31261254 on Build GRCh38
rs753176898 Chr.13: 31260980-31260980 on Build GRCh38
rs759071109 Chr.13: 31260960-31260960 on Build GRCh38
rs759120028 Chr.13: 31261056-31261056 on Build GRCh38
rs187312224 Chr.13: 31261214-31261214 on Build GRCh38
rs756244003 Chr.13: 31261118-31261118 on Build GRCh38
rs759419599 Chr.13: 31261106-31261107 on Build GRCh38
rs367994747 Chr.13: 31261091-31261091 on Build GRCh38
rs922287437 Chr.13: 31261136-31261136 on Build GRCh38
rs751795889 Chr.13: 31260991-31260991 on Build GRCh38
rs764716498 Chr.13: 31261211-31261211 on Build GRCh38
rs770780161 Chr.13: 31261129-31261129 on Build GRCh38
rs745980708 Chr.13: 31261005-31261005 on Build GRCh38
rs527361142 Chr.13: 31261336-31261336 on Build GRCh38
rs559725071 Chr.13: 31261247-31261247 on Build GRCh38
rs781244059 Chr.13: 31260924-31260924 on Build GRCh38
rs756122955 Chr.13: 31261010-31261010 on Build GRCh38
rs758747524 Chr.13: 31260908-31260908 on Build GRCh38
rs1050642275 Chr.13: 31261188-31261188 on Build GRCh38
rs771842620 Chr.13: 31260948-31260948 on Build GRCh38
rs766685203 Chr.13: 31261131-31261132 on Build GRCh38
rs34409451 Chr.13: 31261132-31261132 on Build GRCh38
rs112736590 Chr.13: 31260895-31260895 on Build GRCh38
rs926165449 Chr.13: 31261044-31261044 on Build GRCh38
rs536785414 Chr.13: 31261325-31261325 on Build GRCh38
rs565032858 Chr.13: 31261248-31261248 on Build GRCh38
rs767733375 Chr.13: 31261110-31261110 on Build GRCh38
rs770169224 Chr.13: 31260928-31260929 on Build GRCh38
rs757438335 Chr.13: 31260994-31260994 on Build GRCh38
rs976708734 Chr.13: 31260952-31260952 on Build GRCh38
rs761519440 Chr.13: 31260938-31260938 on Build GRCh38
rs777858836 Chr.13: 31260909-31260909 on Build GRCh38
rs780178206 Chr.13: 31261121-31261121 on Build GRCh38
rs764406422 Chr.13: 31261094-31261094 on Build GRCh38
rs34335590 Chr.13: 31261290-31261290 on Build GRCh38
rs763939190 Chr.13: 31260915-31260915 on Build GRCh38
rs888319226 Chr.13: 31261185-31261185 on Build GRCh38
rs569228688 Chr.13: 31261292-31261292 on Build GRCh38
rs760674958 Chr.13: 31260934-31260934 on Build GRCh38
rs1025436104 Chr.13: 31261326-31261326 on Build GRCh38
rs757567716 Chr.13: 31261106-31261106 on Build GRCh38
rs768556535 Chr.13: 31260933-31260933 on Build GRCh38
rs916293675 Chr.13: 31261072-31261072 on Build GRCh38
rs774233179 Chr.13: 31260937-31260937 on Build GRCh38
rs764779321 Chr.13: 31261065-31261065 on Build GRCh38
rs748104539 Chr.13: 31261012-31261012 on Build GRCh38

View Full Product Details