Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 31284515-31285051 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B3GLCT
Entrez Gene ID
145173
Gene Name
beta 3-glucosyltransferase
Gene Aliases
B3GALTL, B3GTL, B3Glc-T, Gal-T, beta3Glc-T
UniGene
Hs.13205
Gene Chromosome Location
Chr.13: 31199975-31332276 on Build GRCh38
Transcripts
NM_194318.3 XM_006719768.3 XM_011534936.1 XM_011534938.2 XM_017020395.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6240324 c.959A>G p.D320G Chr.13: 31284756-31284756 on Build GRCh38
6609255 c.933T>C p.T311T Chr.13: 31284730-31284730 on Build GRCh38
5037237 c.855T>C p.P285P Chr.13: 31284652-31284652 on Build GRCh38
6074255 c.936G>A p.V312V Chr.13: 31284733-31284733 on Build GRCh38
459299 c.895G>A p.E299K Chr.13: 31284692-31284692 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779159399 Chr.13: 31284765-31284765 on Build GRCh38
rs973201709 Chr.13: 31284825-31284825 on Build GRCh38
rs765006994 Chr.13: 31284675-31284675 on Build GRCh38
rs761254550 Chr.13: 31284948-31284948 on Build GRCh38
rs906937773 Chr.13: 31284703-31284703 on Build GRCh38
rs570447557 Chr.13: 31284565-31284565 on Build GRCh38
rs958563950 Chr.13: 31284936-31284936 on Build GRCh38
rs372993645 Chr.13: 31284746-31284746 on Build GRCh38
rs745686985 Chr.13: 31284651-31284651 on Build GRCh38
rs116754126 Chr.13: 31284653-31284653 on Build GRCh38
rs780534914 Chr.13: 31284821-31284821 on Build GRCh38
rs193177521 Chr.13: 31284784-31284784 on Build GRCh38
rs773628795 Chr.13: 31284645-31284645 on Build GRCh38
rs1045681533 Chr.13: 31284897-31284897 on Build GRCh38
rs1047354805 Chr.13: 31284721-31284721 on Build GRCh38
rs933327901 Chr.13: 31284537-31284537 on Build GRCh38
rs577136281 Chr.13: 31284974-31284974 on Build GRCh38
rs111865672 Chr.13: 31284571-31284571 on Build GRCh38
rs114184584 Chr.13: 31284699-31284699 on Build GRCh38
rs864309632 Chr.13: 31284714-31284714 on Build GRCh38
rs776123474 Chr.13: 31284937-31284937 on Build GRCh38
rs769612244 Chr.13: 31284799-31284799 on Build GRCh38
rs759403205 Chr.13: 31284677-31284685 on Build GRCh38
rs750902054 Chr.13: 31284712-31284712 on Build GRCh38
rs866429355 Chr.13: 31284932-31284932 on Build GRCh38
rs749517667 Chr.13: 31284618-31284618 on Build GRCh38
rs1054296671 Chr.13: 31284929-31284929 on Build GRCh38
rs948501947 Chr.13: 31284692-31284692 on Build GRCh38
rs183305361 Chr.13: 31284907-31284907 on Build GRCh38
rs918550784 Chr.13: 31284541-31284541 on Build GRCh38
rs114996774 Chr.13: 31284978-31284978 on Build GRCh38
rs531369095 Chr.13: 31284620-31284620 on Build GRCh38
rs7993050 Chr.13: 31284902-31284902 on Build GRCh38
rs1030435728 Chr.13: 31284853-31284853 on Build GRCh38
rs771942670 Chr.13: 31284803-31284803 on Build GRCh38
rs775268382 Chr.13: 31284808-31284808 on Build GRCh38
rs752410589 Chr.13: 31284678-31284678 on Build GRCh38
rs554344705 Chr.13: 31284813-31284813 on Build GRCh38
rs113489803 Chr.13: 31284751-31284751 on Build GRCh38
rs375716301 Chr.13: 31284624-31284624 on Build GRCh38
rs781554214 Chr.13: 31284708-31284708 on Build GRCh38
rs866590741 Chr.13: 31284857-31284857 on Build GRCh38
rs761905227 Chr.13: 31284631-31284631 on Build GRCh38
rs770082964 Chr.13: 31284638-31284639 on Build GRCh38
rs377303903 Chr.13: 31284803-31284803 on Build GRCh38
rs147990534 Chr.13: 31284735-31284735 on Build GRCh38
rs769023755 Chr.13: 31284759-31284759 on Build GRCh38
rs950078047 Chr.13: 31284879-31284879 on Build GRCh38
rs543628706 Chr.13: 31284970-31284970 on Build GRCh38
rs868803899 Chr.13: 31284753-31284753 on Build GRCh38
rs948241202 Chr.13: 31285007-31285007 on Build GRCh38
rs950014482 Chr.13: 31284549-31284549 on Build GRCh38
rs771729815 Chr.13: 31284690-31284690 on Build GRCh38
rs748183370 Chr.13: 31284770-31284770 on Build GRCh38
rs759461926 Chr.13: 31284796-31284796 on Build GRCh38
rs1002965291 Chr.13: 31284713-31284713 on Build GRCh38
rs539477123 Chr.13: 31284864-31284864 on Build GRCh38
rs892893561 Chr.13: 31284677-31284677 on Build GRCh38
rs995988715 Chr.13: 31284849-31284849 on Build GRCh38
rs539872859 Chr.13: 31284905-31284905 on Build GRCh38
rs756748782 Chr.13: 31284700-31284700 on Build GRCh38
rs1016134275 Chr.13: 31284809-31284809 on Build GRCh38
rs774275342 Chr.13: 31284630-31284630 on Build GRCh38
rs775240347 Chr.13: 31284802-31284802 on Build GRCh38
rs143454374 Chr.13: 31284688-31284688 on Build GRCh38
rs962778424 Chr.13: 31284822-31284826 on Build GRCh38
rs985592764 Chr.13: 31284812-31284812 on Build GRCh38
rs748241038 Chr.13: 31284613-31284614 on Build GRCh38
rs576826377 Chr.13: 31284938-31284938 on Build GRCh38
rs370709974 Chr.13: 31284637-31284637 on Build GRCh38
rs201027107 Chr.13: 31284729-31284729 on Build GRCh38
rs1041840623 Chr.13: 31285011-31285011 on Build GRCh38

View Full Product Details