Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 66387070-66387562 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAP2K1
Entrez Gene ID
5604
Gene Name
mitogen-activated protein kinase kinase 1
Gene Aliases
CFC3, MAPKK1, MEK1, MKK1, PRKMK1
UniGene
Hs.145442
Gene Chromosome Location
Chr.15: 66386873-66491544 on Build GRCh38
Transcripts
NM_002755.3 XM_017022411.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3816609 c.12G>T p.K4N Chr.15: 66387359-66387359 on Build GRCh38
6555011 c.43C>T p.P15S Chr.15: 66387390-66387390 on Build GRCh38
5768380 c.40G>T p.A14S Chr.15: 66387387-66387387 on Build GRCh38
6953658 c.16C>A p.P6T Chr.15: 66387363-66387363 on Build GRCh38
6931156 c.65G>T p.G22V Chr.15: 66387412-66387412 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs916502006 Chr.15: 66387390-66387390 on Build GRCh38
rs895034214 Chr.15: 66387132-66387132 on Build GRCh38
rs769572694 Chr.15: 66387475-66387475 on Build GRCh38
rs1028000379 Chr.15: 66387182-66387182 on Build GRCh38
rs928727939 Chr.15: 66387526-66387526 on Build GRCh38
rs111281925 Chr.15: 66387205-66387205 on Build GRCh38
rs1052607112 Chr.15: 66387164-66387164 on Build GRCh38
rs571248158 Chr.15: 66387305-66387305 on Build GRCh38
rs574754285 Chr.15: 66387522-66387522 on Build GRCh38
rs983840717 Chr.15: 66387200-66387201 on Build GRCh38
rs886051365 Chr.15: 66387128-66387128 on Build GRCh38
rs77796976 Chr.15: 66387346-66387346 on Build GRCh38
rs71447901 Chr.15: 66387513-66387513 on Build GRCh38
rs761617161 Chr.15: 66387316-66387316 on Build GRCh38
rs1006883752 Chr.15: 66387491-66387491 on Build GRCh38
rs539168006 Chr.15: 66387457-66387457 on Build GRCh38
rs1053188266 Chr.15: 66387279-66387279 on Build GRCh38
rs972542026 Chr.15: 66387329-66387329 on Build GRCh38
rs553162958 Chr.15: 66387421-66387421 on Build GRCh38
rs766163609 Chr.15: 66387454-66387454 on Build GRCh38
rs876657503 Chr.15: 66387386-66387386 on Build GRCh38
rs928678852 Chr.15: 66387336-66387336 on Build GRCh38
rs969880085 Chr.15: 66387174-66387174 on Build GRCh38
rs935578413 Chr.15: 66387509-66387509 on Build GRCh38
rs922139192 Chr.15: 66387109-66387109 on Build GRCh38
rs909365334 Chr.15: 66387267-66387267 on Build GRCh38
rs761150136 Chr.15: 66387358-66387358 on Build GRCh38
rs772853727 Chr.15: 66387341-66387341 on Build GRCh38
rs1019362221 Chr.15: 66387478-66387478 on Build GRCh38
rs530407397 Chr.15: 66387314-66387314 on Build GRCh38
rs561213249 Chr.15: 66387150-66387150 on Build GRCh38
rs377720622 Chr.15: 66387353-66387353 on Build GRCh38
rs998944512 Chr.15: 66387129-66387129 on Build GRCh38
rs41277718 Chr.15: 66387268-66387268 on Build GRCh38
rs1012104352 Chr.15: 66387160-66387160 on Build GRCh38
rs748927101 Chr.15: 66387445-66387446 on Build GRCh38
rs369406498 Chr.15: 66387190-66387190 on Build GRCh38
rs868181833 Chr.15: 66387385-66387385 on Build GRCh38
rs965670100 Chr.15: 66387194-66387194 on Build GRCh38
rs935625030 Chr.15: 66387273-66387273 on Build GRCh38
rs762999277 Chr.15: 66387451-66387451 on Build GRCh38
rs949715211 Chr.15: 66387523-66387523 on Build GRCh38
rs558949572 Chr.15: 66387130-66387130 on Build GRCh38
rs4483802 Chr.15: 66387481-66387481 on Build GRCh38
rs886051364 Chr.15: 66387133-66387134 on Build GRCh38
rs575854200 Chr.15: 66387236-66387237 on Build GRCh38
rs141005341 Chr.15: 66387126-66387127 on Build GRCh38
rs748036330 Chr.15: 66387461-66387461 on Build GRCh38
rs895540023 Chr.15: 66387144-66387144 on Build GRCh38
rs974837747 Chr.15: 66387228-66387229 on Build GRCh38
rs374238278 Chr.15: 66387455-66387455 on Build GRCh38
rs994693514 Chr.15: 66387411-66387411 on Build GRCh38
rs530252769 Chr.15: 66387179-66387179 on Build GRCh38
rs939271011 Chr.15: 66387097-66387097 on Build GRCh38
rs895521497 Chr.15: 66387313-66387313 on Build GRCh38
rs575529090 Chr.15: 66387143-66387143 on Build GRCh38
rs540384307 Chr.15: 66387217-66387217 on Build GRCh38
rs551186099 Chr.15: 66387304-66387304 on Build GRCh38
rs903610094 Chr.15: 66387165-66387165 on Build GRCh38
rs982392486 Chr.15: 66387261-66387279 on Build GRCh38
rs1016458727 Chr.15: 66387495-66387495 on Build GRCh38
rs727504413 Chr.15: 66387403-66387403 on Build GRCh38
rs575611618 Chr.15: 66387480-66387480 on Build GRCh38
rs907198296 Chr.15: 66387096-66387096 on Build GRCh38
rs534948137 Chr.15: 66387228-66387228 on Build GRCh38
rs140749690 Chr.15: 66387416-66387416 on Build GRCh38
rs979995484 Chr.15: 66387502-66387502 on Build GRCh38
rs730880340 Chr.15: 66387317-66387318 on Build GRCh38
rs567535653 Chr.15: 66387410-66387410 on Build GRCh38
rs953592546 Chr.15: 66387261-66387261 on Build GRCh38
rs769505606 Chr.15: 66387331-66387331 on Build GRCh38
rs544924354 Chr.15: 66387148-66387148 on Build GRCh38
rs554848489 Chr.15: 66387507-66387507 on Build GRCh38
rs1044373600 Chr.15: 66387118-66387118 on Build GRCh38
rs764535990 Chr.15: 66387391-66387391 on Build GRCh38
rs968126639 Chr.15: 66387498-66387498 on Build GRCh38
rs1044147889 Chr.15: 66387526-66387527 on Build GRCh38
rs559034006 Chr.15: 66387227-66387227 on Build GRCh38
rs946967890 Chr.15: 66387114-66387114 on Build GRCh38
rs1029051572 Chr.15: 66387484-66387485 on Build GRCh38
rs56149436 Chr.15: 66387460-66387460 on Build GRCh38
rs112542693 Chr.15: 66387263-66387263 on Build GRCh38
rs768750195 Chr.15: 66387310-66387310 on Build GRCh38
rs540517860 Chr.15: 66387529-66387529 on Build GRCh38
rs922065244 Chr.15: 66387234-66387235 on Build GRCh38
rs911926667 Chr.15: 66387206-66387206 on Build GRCh38
rs1022507721 Chr.15: 66387163-66387163 on Build GRCh38

View Full Product Details