Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 1351669-1352167 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNPTG
Entrez Gene ID
84572
Gene Name
N-acetylglucosamine-1-phosphate transferase gamma subunit
Gene Aliases
C16orf27, GNPTAG, LP2537, RJD9
UniGene
Hs.241575
Gene Chromosome Location
Chr.16: 1351899-1363351 on Build GRCh38
Transcripts
NM_032520.4 XM_017023782.1 NM_001001410.2

Gene Symbol
TSR3
Entrez Gene ID
115939
Gene Name
TSR3, acp transferase ribosome maturation factor
Gene Aliases
C16orf42
UniGene
Hs.134846
Gene Chromosome Location
Chr.16: 1349240-1351872 on Build GRCh38
Transcripts
NM_032520.4 XM_017023782.1 NM_001001410.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6349664 c.52C>T p.R18W Chr.16: 1351753-1351753 on Build GRCh38
5657723 c.25G>T p.G9W Chr.16: 1351780-1351780 on Build GRCh38
6436520 c.93G>T p.E31D Chr.16: 1351712-1351712 on Build GRCh38
6390809 c.111G>T p.Q37H Chr.16: 1351694-1351694 on Build GRCh38
6586955 c.110+8G>A p.? Chr.16: 1352167-1352167 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1054163328 Chr.16: 1351759-1351759 on Build GRCh38
rs888194280 Chr.16: 1351777-1351777 on Build GRCh38
rs747247122 Chr.16: 1351995-1351995 on Build GRCh38
rs536513000 Chr.16: 1351731-1351731 on Build GRCh38
rs905778790 Chr.16: 1351918-1351933 on Build GRCh38
rs564853174 Chr.16: 1352092-1352092 on Build GRCh38
rs993454027 Chr.16: 1352036-1352036 on Build GRCh38
rs760140489 Chr.16: 1351765-1351765 on Build GRCh38
rs763764801 Chr.16: 1351974-1351974 on Build GRCh38
rs986694444 Chr.16: 1351955-1351955 on Build GRCh38
rs1016193943 Chr.16: 1351924-1351924 on Build GRCh38
rs8062301 Chr.16: 1351795-1351795 on Build GRCh38
rs547587708 Chr.16: 1351701-1351701 on Build GRCh38
rs1034850148 Chr.16: 1351826-1351826 on Build GRCh38
rs756910244 Chr.16: 1351977-1351977 on Build GRCh38
rs111834368 Chr.16: 1351766-1351766 on Build GRCh38
rs1006329439 Chr.16: 1351922-1351922 on Build GRCh38
rs942360505 Chr.16: 1351744-1351744 on Build GRCh38
rs758954616 Chr.16: 1352107-1352107 on Build GRCh38
rs567583327 Chr.16: 1351703-1351703 on Build GRCh38
rs1041664887 Chr.16: 1351906-1351906 on Build GRCh38
rs8052503 Chr.16: 1352142-1352142 on Build GRCh38
rs532236241 Chr.16: 1351984-1351984 on Build GRCh38
rs972306295 Chr.16: 1351841-1351841 on Build GRCh38
rs562191082 Chr.16: 1351993-1351993 on Build GRCh38
rs1003116954 Chr.16: 1351908-1351908 on Build GRCh38
rs972447741 Chr.16: 1351975-1351975 on Build GRCh38
rs565020861 Chr.16: 1351811-1351811 on Build GRCh38
rs932328410 Chr.16: 1351761-1351761 on Build GRCh38
rs1055666228 Chr.16: 1351911-1351911 on Build GRCh38
rs914153555 Chr.16: 1351732-1351732 on Build GRCh38
rs886051657 Chr.16: 1351924-1351924 on Build GRCh38
rs767184877 Chr.16: 1352094-1352094 on Build GRCh38
rs923463438 Chr.16: 1351714-1351714 on Build GRCh38
rs893881176 Chr.16: 1351927-1351927 on Build GRCh38
rs988271287 Chr.16: 1351724-1351724 on Build GRCh38
rs989859381 Chr.16: 1352115-1352115 on Build GRCh38
rs897789631 Chr.16: 1352028-1352028 on Build GRCh38
rs942495145 Chr.16: 1351874-1351874 on Build GRCh38
rs772239803 Chr.16: 1351991-1351991 on Build GRCh38
rs28541713 Chr.16: 1351844-1351844 on Build GRCh38
rs775363099 Chr.16: 1352075-1352075 on Build GRCh38
rs964111307 Chr.16: 1351960-1351960 on Build GRCh38
rs545117390 Chr.16: 1352073-1352073 on Build GRCh38
rs886051658 Chr.16: 1351971-1351971 on Build GRCh38
rs867217705 Chr.16: 1351973-1351973 on Build GRCh38
rs543660760 Chr.16: 1351978-1351978 on Build GRCh38
rs938573026 Chr.16: 1352148-1352148 on Build GRCh38
rs955019316 Chr.16: 1351949-1351949 on Build GRCh38
rs762469473 Chr.16: 1352077-1352077 on Build GRCh38
rs1021014743 Chr.16: 1351943-1351943 on Build GRCh38
rs771120488 Chr.16: 1352114-1352114 on Build GRCh38
rs1054511796 Chr.16: 1351885-1351885 on Build GRCh38
rs907048726 Chr.16: 1351789-1351789 on Build GRCh38
rs1030320804 Chr.16: 1352037-1352037 on Build GRCh38
rs928528974 Chr.16: 1351988-1351988 on Build GRCh38
rs776466792 Chr.16: 1351743-1351743 on Build GRCh38
rs910957824 Chr.16: 1351869-1351869 on Build GRCh38
rs754166478 Chr.16: 1352098-1352098 on Build GRCh38
rs1041550517 Chr.16: 1351784-1351784 on Build GRCh38
rs575933499 Chr.16: 1352020-1352020 on Build GRCh38
rs928343311 Chr.16: 1352145-1352145 on Build GRCh38
rs945918643 Chr.16: 1351897-1351897 on Build GRCh38
rs771155803 Chr.16: 1352002-1352002 on Build GRCh38
rs967153657 Chr.16: 1352122-1352122 on Build GRCh38
rs755303199 Chr.16: 1352099-1352099 on Build GRCh38
rs1044660992 Chr.16: 1351992-1351992 on Build GRCh38
rs556601696 Chr.16: 1351742-1351742 on Build GRCh38
rs906996362 Chr.16: 1351907-1351907 on Build GRCh38
rs944329193 Chr.16: 1351733-1351733 on Build GRCh38
rs1020962323 Chr.16: 1352066-1352066 on Build GRCh38
rs779505508 Chr.16: 1351985-1351985 on Build GRCh38
rs778042839 Chr.16: 1352110-1352110 on Build GRCh38
rs541284873 Chr.16: 1351926-1351926 on Build GRCh38
rs866920187 Chr.16: 1352104-1352104 on Build GRCh38
rs113470241 Chr.16: 1352021-1352021 on Build GRCh38
rs765220411 Chr.16: 1351938-1351938 on Build GRCh38
rs893047539 Chr.16: 1351760-1351760 on Build GRCh38
rs868174441 Chr.16: 1351825-1351825 on Build GRCh38
rs751036365 Chr.16: 1352086-1352086 on Build GRCh38
rs984755560 Chr.16: 1352140-1352140 on Build GRCh38
rs59225734 Chr.16: 1351929-1351944 on Build GRCh38
rs754225455 Chr.16: 1351982-1351982 on Build GRCh38
rs137853826 Chr.16: 1352123-1352123 on Build GRCh38
rs1036898400 Chr.16: 1351803-1351803 on Build GRCh38
rs539165958 Chr.16: 1351833-1351833 on Build GRCh38
rs763639979 Chr.16: 1352082-1352082 on Build GRCh38
rs1014203471 Chr.16: 1351823-1351823 on Build GRCh38
rs890685456 Chr.16: 1352044-1352044 on Build GRCh38
rs753208354 Chr.16: 1352103-1352103 on Build GRCh38
rs752212692 Chr.16: 1351944-1351944 on Build GRCh38
rs776361173 Chr.16: 1352007-1352007 on Build GRCh38
rs865811325 Chr.16: 1351702-1351702 on Build GRCh38
rs747111791 Chr.16: 1352111-1352111 on Build GRCh38
rs759365145 Chr.16: 1352069-1352069 on Build GRCh38
rs958857563 Chr.16: 1351827-1351827 on Build GRCh38
rs779788991 Chr.16: 1352146-1352146 on Build GRCh38
rs570065824 Chr.16: 1351832-1351832 on Build GRCh38
rs958198845 Chr.16: 1351712-1351712 on Build GRCh38
rs954881938 Chr.16: 1351854-1351854 on Build GRCh38
rs962248757 Chr.16: 1351837-1351837 on Build GRCh38
rs993673873 Chr.16: 1351932-1351932 on Build GRCh38
rs894537198 Chr.16: 1351916-1351916 on Build GRCh38
rs28654239 Chr.16: 1351894-1351894 on Build GRCh38
rs547624231 Chr.16: 1352116-1352116 on Build GRCh38
rs1048108633 Chr.16: 1351769-1351769 on Build GRCh38
rs914207699 Chr.16: 1351972-1351972 on Build GRCh38
rs574801192 Chr.16: 1351976-1351976 on Build GRCh38
rs1009539040 Chr.16: 1352048-1352048 on Build GRCh38
rs1032389243 Chr.16: 1351829-1351829 on Build GRCh38
rs986356876 Chr.16: 1351856-1351856 on Build GRCh38
rs745677704 Chr.16: 1352118-1352118 on Build GRCh38
rs895722963 Chr.16: 1351806-1351806 on Build GRCh38
rs554707396 Chr.16: 1351962-1351962 on Build GRCh38
rs779597308 Chr.16: 1352100-1352100 on Build GRCh38
rs886051656 Chr.16: 1351906-1351906 on Build GRCh38
rs527348189 Chr.16: 1352097-1352097 on Build GRCh38
rs1017797000 Chr.16: 1351956-1351956 on Build GRCh38
rs989913303 Chr.16: 1351971-1351971 on Build GRCh38
rs977124651 Chr.16: 1351986-1351986 on Build GRCh38
rs1051298505 Chr.16: 1351726-1351726 on Build GRCh38
rs1004044382 Chr.16: 1351787-1351787 on Build GRCh38
rs748243075 Chr.16: 1351990-1351990 on Build GRCh38
rs933717447 Chr.16: 1351725-1351725 on Build GRCh38
rs867116818 Chr.16: 1351836-1351836 on Build GRCh38
rs942079497 Chr.16: 1352013-1352013 on Build GRCh38

View Full Product Details