Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 3581062-3581566 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLX4
Entrez Gene ID
84464
Gene Name
SLX4 structure-specific endonuclease subunit
Gene Aliases
BTBD12, FANCP, MUS312
UniGene
Hs.143681
Gene Chromosome Location
Chr.16: 3581181-3611598 on Build GRCh38
Transcripts
NM_032444.2 XM_011522715.2 XM_017023775.1

SNP Detail

SNP ID Chromosome Location
rs566202266 Chr.16: 3581312-3581312 on Build GRCh38
rs1030929863 Chr.16: 3581463-3581463 on Build GRCh38
rs996690421 Chr.16: 3581185-3581185 on Build GRCh38
rs886051973 Chr.16: 3581385-3581385 on Build GRCh38
rs1040916195 Chr.16: 3581152-3581152 on Build GRCh38
rs1032936956 Chr.16: 3581398-3581398 on Build GRCh38
rs950434927 Chr.16: 3581484-3581484 on Build GRCh38
rs910318440 Chr.16: 3581220-3581220 on Build GRCh38
rs71388524 Chr.16: 3581250-3581250 on Build GRCh38
rs966723939 Chr.16: 3581153-3581153 on Build GRCh38
rs908137684 Chr.16: 3581362-3581362 on Build GRCh38
rs903907131 Chr.16: 3581458-3581458 on Build GRCh38
rs886051974 Chr.16: 3581471-3581471 on Build GRCh38
rs1040613124 Chr.16: 3581404-3581404 on Build GRCh38
rs902552965 Chr.16: 3581390-3581390 on Build GRCh38
rs531172700 Chr.16: 3581142-3581142 on Build GRCh38
rs1055138945 Chr.16: 3581102-3581102 on Build GRCh38
rs374659314 Chr.16: 3581330-3581330 on Build GRCh38
rs975839434 Chr.16: 3581290-3581290 on Build GRCh38
rs186844544 Chr.16: 3581105-3581105 on Build GRCh38
rs183371548 Chr.16: 3581315-3581315 on Build GRCh38
rs749836546 Chr.16: 3581258-3581258 on Build GRCh38
rs996352736 Chr.16: 3581443-3581443 on Build GRCh38
rs886051972 Chr.16: 3581192-3581192 on Build GRCh38
rs190865827 Chr.16: 3581119-3581119 on Build GRCh38
rs1013600230 Chr.16: 3581130-3581130 on Build GRCh38
rs775797773 Chr.16: 3581168-3581168 on Build GRCh38
rs1038057735 Chr.16: 3581089-3581089 on Build GRCh38
rs371828836 Chr.16: 3581126-3581126 on Build GRCh38
rs1032926694 Chr.16: 3581169-3581169 on Build GRCh38
rs546194478 Chr.16: 3581464-3581464 on Build GRCh38
rs909480456 Chr.16: 3581508-3581508 on Build GRCh38
rs746220623 Chr.16: 3581143-3581143 on Build GRCh38
rs1023538441 Chr.16: 3581506-3581506 on Build GRCh38
rs916376322 Chr.16: 3581305-3581305 on Build GRCh38
rs367599423 Chr.16: 3581110-3581110 on Build GRCh38
rs944918951 Chr.16: 3581369-3581369 on Build GRCh38
rs960576189 Chr.16: 3581278-3581278 on Build GRCh38
rs776544498 Chr.16: 3581414-3581414 on Build GRCh38
rs893032765 Chr.16: 3581399-3581399 on Build GRCh38
rs984438292 Chr.16: 3581496-3581496 on Build GRCh38
rs115690937 Chr.16: 3581535-3581535 on Build GRCh38
rs1002323213 Chr.16: 3581091-3581091 on Build GRCh38
rs999478976 Chr.16: 3581223-3581223 on Build GRCh38
rs78346491 Chr.16: 3581320-3581320 on Build GRCh38
rs898177842 Chr.16: 3581106-3581106 on Build GRCh38
rs557729822 Chr.16: 3581455-3581455 on Build GRCh38
rs770161158 Chr.16: 3581149-3581149 on Build GRCh38
rs1022367239 Chr.16: 3581211-3581211 on Build GRCh38
rs578011999 Chr.16: 3581460-3581460 on Build GRCh38
rs985930186 Chr.16: 3581215-3581215 on Build GRCh38
rs1042468621 Chr.16: 3581363-3581363 on Build GRCh38
rs1044236747 Chr.16: 3581444-3581444 on Build GRCh38
rs1026059309 Chr.16: 3581474-3581474 on Build GRCh38
rs761469388 Chr.16: 3581446-3581446 on Build GRCh38
rs550987587 Chr.16: 3581217-3581217 on Build GRCh38
rs371879504 Chr.16: 3581531-3581531 on Build GRCh38
rs773005109 Chr.16: 3581505-3581505 on Build GRCh38
rs532555882 Chr.16: 3581148-3581148 on Build GRCh38
rs75773027 Chr.16: 3581338-3581338 on Build GRCh38
rs573496854 Chr.16: 3581182-3581182 on Build GRCh38
rs57699393 Chr.16: 3581358-3581358 on Build GRCh38
rs542189849 Chr.16: 3581516-3581516 on Build GRCh38
rs149634682 Chr.16: 3581307-3581307 on Build GRCh38
rs117958592 Chr.16: 3581301-3581301 on Build GRCh38
rs999212928 Chr.16: 3581156-3581156 on Build GRCh38
rs886652250 Chr.16: 3581144-3581144 on Build GRCh38
rs879150608 Chr.16: 3581454-3581454 on Build GRCh38
rs866821414 Chr.16: 3581141-3581141 on Build GRCh38
rs900698241 Chr.16: 3581412-3581412 on Build GRCh38
rs116498263 Chr.16: 3581247-3581247 on Build GRCh38

View Full Product Details