Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 12655502-12656043 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAN2B1
Entrez Gene ID
4125
Gene Name
mannosidase alpha class 2B member 1
Gene Aliases
LAMAN, MANB
UniGene
Hs.356769
Gene Chromosome Location
Chr.19: 12646508-12666777 on Build GRCh38
Transcripts
NM_000528.3 NM_001173498.1 XM_005259913.1 XM_017026818.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
341123 c.1686G>T p.E562D Chr.19: 12655835-12655835 on Build GRCh38
5981453 c.1665C>A p.D555E Chr.19: 12655856-12655856 on Build GRCh38
1193855 c.1697C>T p.S566L Chr.19: 12655824-12655824 on Build GRCh38
5984193 c.1703C>G p.S568* Chr.19: 12655818-12655818 on Build GRCh38
4951378 c.1802C>T p.S601F Chr.19: 12655719-12655719 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs548270743 Chr.19: 12655668-12655668 on Build GRCh38
rs773178744 Chr.19: 12655838-12655838 on Build GRCh38
rs181476648 Chr.19: 12655728-12655728 on Build GRCh38
rs367980012 Chr.19: 12655805-12655805 on Build GRCh38
rs376719984 Chr.19: 12655860-12655860 on Build GRCh38
rs769653813 Chr.19: 12655737-12655737 on Build GRCh38
rs757635212 Chr.19: 12655784-12655784 on Build GRCh38
rs540409826 Chr.19: 12655725-12655725 on Build GRCh38
rs531106814 Chr.19: 12655990-12655990 on Build GRCh38
rs368245289 Chr.19: 12655839-12655839 on Build GRCh38
rs750784245 Chr.19: 12655799-12655799 on Build GRCh38
rs1002782013 Chr.19: 12655575-12655575 on Build GRCh38
rs777277411 Chr.19: 12655879-12655879 on Build GRCh38
rs995344408 Chr.19: 12655981-12655981 on Build GRCh38
rs755140505 Chr.19: 12655862-12655862 on Build GRCh38
rs996496763 Chr.19: 12655859-12655859 on Build GRCh38
rs1057516927 Chr.19: 12655837-12655837 on Build GRCh38
rs762375401 Chr.19: 12655847-12655847 on Build GRCh38
rs1057516459 Chr.19: 12655741-12655750 on Build GRCh38
rs59357922 Chr.19: 12655780-12655780 on Build GRCh38
rs138349480 Chr.19: 12655850-12655850 on Build GRCh38
rs759033353 Chr.19: 12655769-12655769 on Build GRCh38
rs746448122 Chr.19: 12655855-12655855 on Build GRCh38
rs756166631 Chr.19: 12655891-12655891 on Build GRCh38
rs772270721 Chr.19: 12655716-12655716 on Build GRCh38
rs761703995 Chr.19: 12655734-12655734 on Build GRCh38
rs398123454 Chr.19: 12655816-12655817 on Build GRCh38
rs956909161 Chr.19: 12655747-12655747 on Build GRCh38
rs749064347 Chr.19: 12655901-12655901 on Build GRCh38
rs765862201 Chr.19: 12655659-12655659 on Build GRCh38
rs374506711 Chr.19: 12655803-12655803 on Build GRCh38
rs912714124 Chr.19: 12655753-12655753 on Build GRCh38
rs371071601 Chr.19: 12655726-12655726 on Build GRCh38
rs777404968 Chr.19: 12656010-12656010 on Build GRCh38
rs891030696 Chr.19: 12655881-12655881 on Build GRCh38
rs887029029 Chr.19: 12655559-12655559 on Build GRCh38
rs766998624 Chr.19: 12655843-12655843 on Build GRCh38
rs774970404 Chr.19: 12655845-12655845 on Build GRCh38
rs369999433 Chr.19: 12655893-12655893 on Build GRCh38
rs773893054 Chr.19: 12655912-12655912 on Build GRCh38
rs80338677 Chr.19: 12655693-12655693 on Build GRCh38
rs769847219 Chr.19: 12655831-12655831 on Build GRCh38
rs748152945 Chr.19: 12655829-12655829 on Build GRCh38
rs762564359 Chr.19: 12655652-12655652 on Build GRCh38
rs552429021 Chr.19: 12655931-12655931 on Build GRCh38
rs150479159 Chr.19: 12655816-12655816 on Build GRCh38
rs746035776 Chr.19: 12655706-12655706 on Build GRCh38
rs375721133 Chr.19: 12655915-12655915 on Build GRCh38
rs557827242 Chr.19: 12655614-12655614 on Build GRCh38
rs8107642 Chr.19: 12655556-12655556 on Build GRCh38
rs1052563225 Chr.19: 12655532-12655532 on Build GRCh38
rs376163192 Chr.19: 12655691-12655691 on Build GRCh38
rs868050025 Chr.19: 12655866-12655866 on Build GRCh38
rs372645746 Chr.19: 12655906-12655906 on Build GRCh38
rs766513928 Chr.19: 12655856-12655856 on Build GRCh38
rs751073488 Chr.19: 12655674-12655674 on Build GRCh38
rs987054644 Chr.19: 12655751-12655751 on Build GRCh38
rs930982072 Chr.19: 12655536-12655536 on Build GRCh38
rs377104016 Chr.19: 12655810-12655810 on Build GRCh38
rs745594759 Chr.19: 12655924-12655924 on Build GRCh38
rs969379236 Chr.19: 12655736-12655736 on Build GRCh38
rs1015430702 Chr.19: 12655976-12655976 on Build GRCh38
rs1000868549 Chr.19: 12655738-12655738 on Build GRCh38
rs765651082 Chr.19: 12655793-12655793 on Build GRCh38
rs771628343 Chr.19: 12655861-12655861 on Build GRCh38
rs373101689 Chr.19: 12655864-12655864 on Build GRCh38
rs199588220 Chr.19: 12655848-12655848 on Build GRCh38
rs764543590 Chr.19: 12655771-12655771 on Build GRCh38
rs747081586 Chr.19: 12655811-12655811 on Build GRCh38
rs1027707932 Chr.19: 12656004-12656004 on Build GRCh38
rs146778702 Chr.19: 12655745-12655745 on Build GRCh38
rs962577495 Chr.19: 12655977-12655977 on Build GRCh38
rs544475124 Chr.19: 12655752-12655752 on Build GRCh38
rs145062583 Chr.19: 12655702-12655702 on Build GRCh38
rs766021102 Chr.19: 12655750-12655750 on Build GRCh38
rs771580613 Chr.19: 12655613-12655613 on Build GRCh38
rs899530842 Chr.19: 12655595-12655595 on Build GRCh38
rs938576591 Chr.19: 12655880-12655880 on Build GRCh38
rs1012683096 Chr.19: 12655964-12655964 on Build GRCh38
rs864621986 Chr.19: 12655830-12655830 on Build GRCh38
rs781005116 Chr.19: 12655820-12655820 on Build GRCh38
rs773628674 Chr.19: 12655932-12655932 on Build GRCh38
rs903927998 Chr.19: 12656011-12656011 on Build GRCh38
rs573996230 Chr.19: 12655742-12655742 on Build GRCh38
rs748206062 Chr.19: 12655882-12655882 on Build GRCh38

View Full Product Details