Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 18786156-18786661 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COMP
Entrez Gene ID
1311
Gene Name
cartilage oligomeric matrix protein
Gene Aliases
EDM1, EPD1, MED, PSACH, THBS5, TSP5
UniGene
Hs.1584
Gene Chromosome Location
Chr.19: 18782773-18791305 on Build GRCh38
Transcripts
NM_000095.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1680808 c.1180G>T p.D394Y Chr.19: 18786606-18786606 on Build GRCh38
5921406 c.1136G>A p.R379Q Chr.19: 18786650-18786650 on Build GRCh38
3672795 c.1147C>G p.Q383E Chr.19: 18786639-18786639 on Build GRCh38
711091 c.1248G>A p.P416P Chr.19: 18786538-18786538 on Build GRCh38
160155 c.1303G>C p.D435H Chr.19: 18786243-18786243 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768654756 Chr.19: 18786216-18786216 on Build GRCh38
rs764879634 Chr.19: 18786534-18786534 on Build GRCh38
rs147630181 Chr.19: 18786353-18786353 on Build GRCh38
rs909891604 Chr.19: 18786463-18786463 on Build GRCh38
rs746578492 Chr.19: 18786520-18786520 on Build GRCh38
rs886042932 Chr.19: 18786564-18786564 on Build GRCh38
rs1007491344 Chr.19: 18786366-18786366 on Build GRCh38
rs776212021 Chr.19: 18786289-18786289 on Build GRCh38
rs771841538 Chr.19: 18786506-18786506 on Build GRCh38
rs200382113 Chr.19: 18786489-18786489 on Build GRCh38
rs201063220 Chr.19: 18786586-18786586 on Build GRCh38
rs781064481 Chr.19: 18786330-18786330 on Build GRCh38
rs796435646 Chr.19: 18786423-18786423 on Build GRCh38
rs1041454680 Chr.19: 18786310-18786310 on Build GRCh38
rs745555403 Chr.19: 18786623-18786623 on Build GRCh38
rs887239135 Chr.19: 18786523-18786523 on Build GRCh38
rs779078342 Chr.19: 18786192-18786192 on Build GRCh38
rs528720862 Chr.19: 18786358-18786358 on Build GRCh38
rs765963605 Chr.19: 18786180-18786180 on Build GRCh38
rs753124522 Chr.19: 18786201-18786201 on Build GRCh38
rs747972155 Chr.19: 18786221-18786221 on Build GRCh38
rs764599385 Chr.19: 18786311-18786311 on Build GRCh38
rs984081635 Chr.19: 18786456-18786456 on Build GRCh38
rs775203006 Chr.19: 18786634-18786634 on Build GRCh38
rs754834870 Chr.19: 18786496-18786496 on Build GRCh38
rs968351328 Chr.19: 18786345-18786345 on Build GRCh38
rs201124517 Chr.19: 18786296-18786296 on Build GRCh38
rs751860122 Chr.19: 18786540-18786540 on Build GRCh38
rs755577871 Chr.19: 18786187-18786187 on Build GRCh38
rs750654646 Chr.19: 18786562-18786562 on Build GRCh38
rs751814855 Chr.19: 18786313-18786313 on Build GRCh38
rs902975311 Chr.19: 18786327-18786327 on Build GRCh38
rs140712407 Chr.19: 18786370-18786370 on Build GRCh38
rs150080950 Chr.19: 18786401-18786401 on Build GRCh38
rs1032644784 Chr.19: 18786347-18786347 on Build GRCh38
rs778795530 Chr.19: 18786499-18786499 on Build GRCh38
rs557014004 Chr.19: 18786553-18786553 on Build GRCh38
rs902903169 Chr.19: 18786637-18786637 on Build GRCh38
rs758804491 Chr.19: 18786309-18786309 on Build GRCh38
rs749174754 Chr.19: 18786214-18786214 on Build GRCh38
rs748137604 Chr.19: 18786501-18786501 on Build GRCh38
rs61739916 Chr.19: 18786630-18786630 on Build GRCh38
rs537701126 Chr.19: 18786419-18786419 on Build GRCh38
rs371717961 Chr.19: 18786307-18786307 on Build GRCh38
rs746841380 Chr.19: 18786594-18786594 on Build GRCh38
rs764546396 Chr.19: 18786539-18786539 on Build GRCh38
rs368587069 Chr.19: 18786514-18786514 on Build GRCh38
rs368715216 Chr.19: 18786538-18786538 on Build GRCh38
rs200422116 Chr.19: 18786588-18786588 on Build GRCh38
rs754289056 Chr.19: 18786181-18786181 on Build GRCh38
rs770628352 Chr.19: 18786524-18786524 on Build GRCh38
rs763141191 Chr.19: 18786216-18786216 on Build GRCh38
rs770889743 Chr.19: 18786211-18786211 on Build GRCh38
rs375908187 Chr.19: 18786532-18786532 on Build GRCh38
rs765804127 Chr.19: 18786260-18786260 on Build GRCh38
rs767814536 Chr.19: 18786560-18786560 on Build GRCh38
rs972333523 Chr.19: 18786411-18786411 on Build GRCh38
rs770449845 Chr.19: 18786601-18786601 on Build GRCh38
rs1047499216 Chr.19: 18786516-18786516 on Build GRCh38
rs758338295 Chr.19: 18786587-18786587 on Build GRCh38
rs763339761 Chr.19: 18786363-18786363 on Build GRCh38
rs758644769 Chr.19: 18786204-18786204 on Build GRCh38
rs996754896 Chr.19: 18786290-18786290 on Build GRCh38
rs756200063 Chr.19: 18786569-18786569 on Build GRCh38
rs375203531 Chr.19: 18786212-18786212 on Build GRCh38
rs151050419 Chr.19: 18786618-18786618 on Build GRCh38
rs774086015 Chr.19: 18786217-18786217 on Build GRCh38
rs563307476 Chr.19: 18786209-18786209 on Build GRCh38
rs368992893 Chr.19: 18786346-18786347 on Build GRCh38
rs763485657 Chr.19: 18786639-18786639 on Build GRCh38
rs931273850 Chr.19: 18786437-18786437 on Build GRCh38
rs757563215 Chr.19: 18786314-18786314 on Build GRCh38
rs745984596 Chr.19: 18786490-18786490 on Build GRCh38
rs397515510 Chr.19: 18786628-18786630 on Build GRCh38
rs764459421 Chr.19: 18786220-18786220 on Build GRCh38
rs765795221 Chr.19: 18786300-18786300 on Build GRCh38
rs760435349 Chr.19: 18786231-18786231 on Build GRCh38
rs878853098 Chr.19: 18786255-18786267 on Build GRCh38
rs764590130 Chr.19: 18786537-18786537 on Build GRCh38
rs747270281 Chr.19: 18786210-18786210 on Build GRCh38
rs752143454 Chr.19: 18786323-18786326 on Build GRCh38
rs780165445 Chr.19: 18786570-18786570 on Build GRCh38
rs772829625 Chr.19: 18786227-18786227 on Build GRCh38
rs775953172 Chr.19: 18786527-18786527 on Build GRCh38

View Full Product Details