Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 45527705-45528215 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OPA3
Entrez Gene ID
80207
Gene Name
optic atrophy 3 (autosomal recessive, with chorea and spastic paraplegia)
Gene Aliases
MGA3
UniGene
Hs.466945
Gene Chromosome Location
Chr.19: 45527767-45584864 on Build GRCh38
Transcripts
NM_001017989.2

SNP Detail

SNP ID Chromosome Location
rs1053708911 Chr.19: 45527951-45527951 on Build GRCh38
rs757166355 Chr.19: 45527781-45527781 on Build GRCh38
rs982680443 Chr.19: 45527762-45527762 on Build GRCh38
rs960066929 Chr.19: 45528173-45528173 on Build GRCh38
rs1043731753 Chr.19: 45528135-45528135 on Build GRCh38
rs142545664 Chr.19: 45527816-45527816 on Build GRCh38
rs751517859 Chr.19: 45527742-45527742 on Build GRCh38
rs1051562345 Chr.19: 45527853-45527853 on Build GRCh38
rs938421008 Chr.19: 45527935-45527935 on Build GRCh38
rs1002455355 Chr.19: 45528150-45528150 on Build GRCh38
rs150986093 Chr.19: 45527919-45527919 on Build GRCh38
rs984665058 Chr.19: 45527887-45527887 on Build GRCh38
rs913964332 Chr.19: 45528077-45528078 on Build GRCh38
rs1026566779 Chr.19: 45528024-45528024 on Build GRCh38
rs563527167 Chr.19: 45527774-45527774 on Build GRCh38
rs1006175176 Chr.19: 45528053-45528053 on Build GRCh38
rs1055733582 Chr.19: 45528165-45528165 on Build GRCh38
rs941204598 Chr.19: 45527776-45527776 on Build GRCh38
rs751646789 Chr.19: 45527796-45527796 on Build GRCh38
rs748979866 Chr.19: 45528181-45528181 on Build GRCh38
rs779667115 Chr.19: 45528115-45528115 on Build GRCh38
rs950527020 Chr.19: 45528040-45528040 on Build GRCh38
rs534811674 Chr.19: 45527856-45527856 on Build GRCh38
rs928292500 Chr.19: 45528141-45528141 on Build GRCh38
rs907401272 Chr.19: 45527733-45527733 on Build GRCh38
rs186186357 Chr.19: 45528099-45528099 on Build GRCh38
rs889819932 Chr.19: 45527968-45527968 on Build GRCh38
rs934071465 Chr.19: 45527846-45527846 on Build GRCh38
rs913369240 Chr.19: 45527955-45527955 on Build GRCh38
rs947642891 Chr.19: 45527957-45527957 on Build GRCh38
rs750164782 Chr.19: 45528052-45528052 on Build GRCh38
rs965936638 Chr.19: 45527864-45527864 on Build GRCh38
rs901273782 Chr.19: 45528013-45528013 on Build GRCh38
rs948358680 Chr.19: 45527905-45527905 on Build GRCh38
rs908916475 Chr.19: 45527903-45527903 on Build GRCh38
rs1051190717 Chr.19: 45528031-45528031 on Build GRCh38
rs950618307 Chr.19: 45527886-45527886 on Build GRCh38
rs951020638 Chr.19: 45527740-45527740 on Build GRCh38
rs909761793 Chr.19: 45527775-45527775 on Build GRCh38
rs1014149183 Chr.19: 45528189-45528189 on Build GRCh38
rs1044005559 Chr.19: 45527877-45527877 on Build GRCh38
rs755874224 Chr.19: 45528056-45528056 on Build GRCh38
rs781432805 Chr.19: 45528006-45528009 on Build GRCh38
rs548446096 Chr.19: 45527880-45527880 on Build GRCh38
rs1043287822 Chr.19: 45527960-45527960 on Build GRCh38
rs909833518 Chr.19: 45527992-45527992 on Build GRCh38
rs34997904 Chr.19: 45528096-45528097 on Build GRCh38
rs980193512 Chr.19: 45528128-45528128 on Build GRCh38
rs1009548567 Chr.19: 45527873-45527873 on Build GRCh38
rs892481024 Chr.19: 45527871-45527871 on Build GRCh38
rs115623295 Chr.19: 45528014-45528014 on Build GRCh38
rs565799330 Chr.19: 45527854-45527854 on Build GRCh38
rs889746301 Chr.19: 45527815-45527815 on Build GRCh38
rs2270199 Chr.19: 45527807-45527807 on Build GRCh38
rs539475374 Chr.19: 45528098-45528098 on Build GRCh38
rs557682873 Chr.19: 45528005-45528005 on Build GRCh38
rs1002400848 Chr.19: 45527892-45527892 on Build GRCh38
rs1024437563 Chr.19: 45527946-45527946 on Build GRCh38
rs928355116 Chr.19: 45527934-45527934 on Build GRCh38
rs1021588844 Chr.19: 45527810-45527810 on Build GRCh38
rs938834263 Chr.19: 45527737-45527737 on Build GRCh38
rs976304761 Chr.19: 45527870-45527870 on Build GRCh38
rs537335238 Chr.19: 45527977-45527977 on Build GRCh38

View Full Product Details