Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 9583947-9584484 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
YWHAQ
Entrez Gene ID
10971
Gene Name
tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein theta
Gene Aliases
14-3-3, 1C5, HS1
UniGene
Hs.74405
Gene Chromosome Location
Chr.2: 9583967-9631055 on Build GRCh38
Transcripts
NM_006826.3

SNP Detail

SNP ID Chromosome Location
rs916117871 Chr.2: 9583977-9583977 on Build GRCh38
rs937089323 Chr.2: 9583978-9583978 on Build GRCh38
rs1057230798 Chr.2: 9583980-9583980 on Build GRCh38
rs537163310 Chr.2: 9584456-9584456 on Build GRCh38
rs1010244282 Chr.2: 9584260-9584260 on Build GRCh38
rs561007341 Chr.2: 9584016-9584016 on Build GRCh38
rs1022589377 Chr.2: 9584261-9584261 on Build GRCh38
rs534763265 Chr.2: 9584374-9584374 on Build GRCh38
rs951353438 Chr.2: 9583973-9583973 on Build GRCh38
rs563513298 Chr.2: 9584153-9584153 on Build GRCh38
rs753719464 Chr.2: 9584077-9584078 on Build GRCh38
rs774661836 Chr.2: 9584444-9584444 on Build GRCh38
rs541231706 Chr.2: 9583988-9583988 on Build GRCh38
rs552177354 Chr.2: 9584223-9584223 on Build GRCh38
rs568125543 Chr.2: 9584382-9584382 on Build GRCh38
rs1036197806 Chr.2: 9584032-9584032 on Build GRCh38
rs899011428 Chr.2: 9584033-9584033 on Build GRCh38
rs980960095 Chr.2: 9584330-9584330 on Build GRCh38
rs548411732 Chr.2: 9584379-9584379 on Build GRCh38
rs532571933 Chr.2: 9584168-9584168 on Build GRCh38
rs994661633 Chr.2: 9584112-9584112 on Build GRCh38
rs956706614 Chr.2: 9584222-9584222 on Build GRCh38
rs866115547 Chr.2: 9584152-9584152 on Build GRCh38
rs916894072 Chr.2: 9584005-9584005 on Build GRCh38
rs6758214 Chr.2: 9584347-9584347 on Build GRCh38
rs931571465 Chr.2: 9583979-9583979 on Build GRCh38
rs1008968417 Chr.2: 9584156-9584156 on Build GRCh38
rs7594 Chr.2: 9584138-9584138 on Build GRCh38
rs540708954 Chr.2: 9584267-9584267 on Build GRCh38
rs940622385 Chr.2: 9584015-9584015 on Build GRCh38
rs757592569 Chr.2: 9584130-9584136 on Build GRCh38
rs868059430 Chr.2: 9584224-9584224 on Build GRCh38
rs187680337 Chr.2: 9584143-9584143 on Build GRCh38
rs968289098 Chr.2: 9584284-9584288 on Build GRCh38
rs191107303 Chr.2: 9584413-9584413 on Build GRCh38

View Full Product Details