Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27366668-27367169 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF2B4
Entrez Gene ID
8890
Gene Name
eukaryotic translation initiation factor 2B subunit delta
Gene Aliases
EIF-2B, EIF2B, EIF2Bdelta
UniGene
Hs.169474
Gene Chromosome Location
Chr.2: 27364352-27370457 on Build GRCh38
Transcripts
NM_001034116.1 NM_001318965.1 NM_001318966.1 NM_001318967.1 NM_001318968.1 NM_001318969.1 NM_015636.3 NM_172195.3 XM_006712132.1 XM_011533147.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3798911 c.1021C>A p.L341M Chr.2: 27366929-27366929 on Build GRCh38
6866954 c.1226C>T p.P409L Chr.2: 27366784-27366784 on Build GRCh38
2939881 c.1091G>A p.R364Q Chr.2: 27366919-27366919 on Build GRCh38
6652722 c.1090C>T p.R364* Chr.2: 27366920-27366920 on Build GRCh38
3734188 c.1094C>G p.P365R Chr.2: 27366856-27366856 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756150677 Chr.2: 27366785-27366785 on Build GRCh38
rs769848938 Chr.2: 27366940-27366940 on Build GRCh38
rs771240893 Chr.2: 27366811-27366811 on Build GRCh38
rs762547412 Chr.2: 27366951-27366953 on Build GRCh38
rs113994033 Chr.2: 27366880-27366880 on Build GRCh38
rs746687079 Chr.2: 27367106-27367106 on Build GRCh38
rs773304003 Chr.2: 27367133-27367133 on Build GRCh38
rs201216936 Chr.2: 27366862-27366862 on Build GRCh38
rs1052771340 Chr.2: 27366967-27366967 on Build GRCh38
rs201674537 Chr.2: 27367058-27367058 on Build GRCh38
rs914273202 Chr.2: 27367017-27367017 on Build GRCh38
rs768087185 Chr.2: 27366926-27366926 on Build GRCh38
rs375697618 Chr.2: 27366869-27366869 on Build GRCh38
rs1050905285 Chr.2: 27367004-27367004 on Build GRCh38
rs768160292 Chr.2: 27366848-27366848 on Build GRCh38
rs772714592 Chr.2: 27366715-27366716 on Build GRCh38
rs750884435 Chr.2: 27367038-27367038 on Build GRCh38
rs140046653 Chr.2: 27366746-27366746 on Build GRCh38
rs113994036 Chr.2: 27366778-27366778 on Build GRCh38
rs113994032 Chr.2: 27366881-27366881 on Build GRCh38
rs745579285 Chr.2: 27366821-27366821 on Build GRCh38
rs2280737 Chr.2: 27366943-27366943 on Build GRCh38
rs767857138 Chr.2: 27366878-27366878 on Build GRCh38
rs765038824 Chr.2: 27366756-27366756 on Build GRCh38
rs555838369 Chr.2: 27366740-27366740 on Build GRCh38
rs201869672 Chr.2: 27366747-27366747 on Build GRCh38
rs867769628 Chr.2: 27366987-27366987 on Build GRCh38
rs749208137 Chr.2: 27367136-27367136 on Build GRCh38
rs113994037 Chr.2: 27366758-27366758 on Build GRCh38
rs771710835 Chr.2: 27366826-27366826 on Build GRCh38
rs184607650 Chr.2: 27367140-27367140 on Build GRCh38
rs144124145 Chr.2: 27366795-27366795 on Build GRCh38
rs758766436 Chr.2: 27366726-27366726 on Build GRCh38
rs947114920 Chr.2: 27367022-27367022 on Build GRCh38
rs536766043 Chr.2: 27367066-27367066 on Build GRCh38
rs113994034 Chr.2: 27366859-27366859 on Build GRCh38
rs555801578 Chr.2: 27366742-27366742 on Build GRCh38
rs754354365 Chr.2: 27366882-27366882 on Build GRCh38
rs776461313 Chr.2: 27366855-27366855 on Build GRCh38
rs370347329 Chr.2: 27366719-27366719 on Build GRCh38
rs766227499 Chr.2: 27366948-27366948 on Build GRCh38
rs779707712 Chr.2: 27366898-27366898 on Build GRCh38
rs755199535 Chr.2: 27366972-27366972 on Build GRCh38
rs141025043 Chr.2: 27366919-27366919 on Build GRCh38
rs757219973 Chr.2: 27366985-27366985 on Build GRCh38
rs950114490 Chr.2: 27366887-27366887 on Build GRCh38
rs760929188 Chr.2: 27366953-27366953 on Build GRCh38
rs886055901 Chr.2: 27366814-27366814 on Build GRCh38
rs746628836 Chr.2: 27366900-27366900 on Build GRCh38
rs760119905 Chr.2: 27366839-27366839 on Build GRCh38
rs780633076 Chr.2: 27367115-27367115 on Build GRCh38
rs890861166 Chr.2: 27367008-27367008 on Build GRCh38
rs749762984 Chr.2: 27366807-27366807 on Build GRCh38
rs553331679 Chr.2: 27366753-27366753 on Build GRCh38
rs781357939 Chr.2: 27367010-27367010 on Build GRCh38
rs764140570 Chr.2: 27366963-27366963 on Build GRCh38
rs765749379 Chr.2: 27367027-27367027 on Build GRCh38
rs766010614 Chr.2: 27366956-27366956 on Build GRCh38
rs113994035 Chr.2: 27366830-27366830 on Build GRCh38
rs368168999 Chr.2: 27366889-27366889 on Build GRCh38
rs779319688 Chr.2: 27366890-27366890 on Build GRCh38
rs200954953 Chr.2: 27367126-27367126 on Build GRCh38
rs755225054 Chr.2: 27366769-27366769 on Build GRCh38
rs751867352 Chr.2: 27366757-27366757 on Build GRCh38
rs144016401 Chr.2: 27366909-27366909 on Build GRCh38
rs770898806 Chr.2: 27367138-27367138 on Build GRCh38
rs773856405 Chr.2: 27366972-27366975 on Build GRCh38
rs917281842 Chr.2: 27366863-27366863 on Build GRCh38
rs371946487 Chr.2: 27366829-27366829 on Build GRCh38
rs199942945 Chr.2: 27366725-27366725 on Build GRCh38
rs775060569 Chr.2: 27366834-27366834 on Build GRCh38
rs145468921 Chr.2: 27366929-27366929 on Build GRCh38
rs754642030 Chr.2: 27367107-27367107 on Build GRCh38
rs760676299 Chr.2: 27366739-27366739 on Build GRCh38
rs935923177 Chr.2: 27366933-27366933 on Build GRCh38
rs753949012 Chr.2: 27366982-27366982 on Build GRCh38
rs779229365 Chr.2: 27366816-27366816 on Build GRCh38
rs769371305 Chr.2: 27367132-27367132 on Build GRCh38
rs567846751 Chr.2: 27366695-27366695 on Build GRCh38
rs778375725 Chr.2: 27366893-27366893 on Build GRCh38
rs369084409 Chr.2: 27366946-27366946 on Build GRCh38

View Full Product Details