Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 41745420-41745914 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PHOX2B
Entrez Gene ID
8929
Gene Name
paired like homeobox 2b
Gene Aliases
NBLST2, NBPhox, PMX2B
UniGene
Hs.87202
Gene Chromosome Location
Chr.4: 41744082-41748970 on Build GRCh38
Transcripts
NM_003924.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1220474 c.851C>T p.P284L Chr.4: 41745901-41745901 on Build GRCh38
7383808 c.925G>C p.V309L Chr.4: 41745827-41745827 on Build GRCh38
5449994 c.919G>A p.A307T Chr.4: 41745833-41745833 on Build GRCh38
6401198 c.904A>G p.N302D Chr.4: 41745848-41745848 on Build GRCh38
7343365 c.875_876CC>TT p.A292V Chr.4: 41745876-41745877 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774951337 Chr.4: 41745774-41745775 on Build GRCh38
rs771616235 Chr.4: 41745775-41745783 on Build GRCh38
rs987008229 Chr.4: 41745561-41745561 on Build GRCh38
rs999505638 Chr.4: 41745689-41745689 on Build GRCh38
rs994949118 Chr.4: 41745593-41745593 on Build GRCh38
rs970155283 Chr.4: 41745493-41745493 on Build GRCh38
rs750728938 Chr.4: 41745825-41745825 on Build GRCh38
rs754671744 Chr.4: 41745769-41745769 on Build GRCh38
rs989465234 Chr.4: 41745541-41745541 on Build GRCh38
rs1047417474 Chr.4: 41745640-41745640 on Build GRCh38
rs780019991 Chr.4: 41745766-41745766 on Build GRCh38
rs759556353 Chr.4: 41745790-41745790 on Build GRCh38
rs768798757 Chr.4: 41745784-41745784 on Build GRCh38
rs779171868 Chr.4: 41745787-41745787 on Build GRCh38
rs186778106 Chr.4: 41745613-41745613 on Build GRCh38
rs749346904 Chr.4: 41745779-41745779 on Build GRCh38
rs912449909 Chr.4: 41745533-41745533 on Build GRCh38
rs762234006 Chr.4: 41745820-41745820 on Build GRCh38
rs886059413 Chr.4: 41745749-41745749 on Build GRCh38
rs769663483 Chr.4: 41745887-41745887 on Build GRCh38
rs114290493 Chr.4: 41745646-41745646 on Build GRCh38
rs75913938 Chr.4: 41745727-41745727 on Build GRCh38
rs755070000 Chr.4: 41745836-41745836 on Build GRCh38
rs898312252 Chr.4: 41745582-41745582 on Build GRCh38
rs763708823 Chr.4: 41745518-41745518 on Build GRCh38
rs538080267 Chr.4: 41745733-41745733 on Build GRCh38
rs779068107 Chr.4: 41745847-41745847 on Build GRCh38
rs989877179 Chr.4: 41745738-41745738 on Build GRCh38
rs772358257 Chr.4: 41745872-41745872 on Build GRCh38
rs939854567 Chr.4: 41745537-41745537 on Build GRCh38
rs769726959 Chr.4: 41745793-41745793 on Build GRCh38
rs775478414 Chr.4: 41745892-41745892 on Build GRCh38
rs774897796 Chr.4: 41745798-41745798 on Build GRCh38
rs756546457 Chr.4: 41745765-41745765 on Build GRCh38
rs753912150 Chr.4: 41745834-41745834 on Build GRCh38
rs551679079 Chr.4: 41745480-41745480 on Build GRCh38
rs1038094061 Chr.4: 41745609-41745609 on Build GRCh38
rs895428574 Chr.4: 41745742-41745761 on Build GRCh38
rs955624967 Chr.4: 41745750-41745750 on Build GRCh38
rs987470852 Chr.4: 41745754-41745754 on Build GRCh38
rs920954240 Chr.4: 41745512-41745512 on Build GRCh38
rs1024985420 Chr.4: 41745629-41745629 on Build GRCh38
rs1025565125 Chr.4: 41745465-41745465 on Build GRCh38
rs934137643 Chr.4: 41745555-41745555 on Build GRCh38
rs748808912 Chr.4: 41745893-41745893 on Build GRCh38
rs748382088 Chr.4: 41745867-41745867 on Build GRCh38
rs977582808 Chr.4: 41745475-41745475 on Build GRCh38
rs973793027 Chr.4: 41745775-41745780 on Build GRCh38
rs756644504 Chr.4: 41745827-41745827 on Build GRCh38
rs770841700 Chr.4: 41745788-41745788 on Build GRCh38
rs17885864 Chr.4: 41745882-41745882 on Build GRCh38
rs780578509 Chr.4: 41745833-41745833 on Build GRCh38
rs913518069 Chr.4: 41745546-41745546 on Build GRCh38
rs1013819064 Chr.4: 41745660-41745660 on Build GRCh38
rs911406786 Chr.4: 41745582-41745585 on Build GRCh38
rs894496721 Chr.4: 41745611-41745611 on Build GRCh38
rs905041383 Chr.4: 41745703-41745703 on Build GRCh38
rs1049154964 Chr.4: 41745607-41745607 on Build GRCh38
rs971984898 Chr.4: 41745445-41745445 on Build GRCh38
rs578163582 Chr.4: 41745753-41745753 on Build GRCh38
rs751212535 Chr.4: 41745810-41745810 on Build GRCh38
rs112714631 Chr.4: 41745542-41745542 on Build GRCh38
rs989845912 Chr.4: 41745489-41745489 on Build GRCh38
rs763704619 Chr.4: 41745802-41745802 on Build GRCh38
rs73810341 Chr.4: 41745529-41745529 on Build GRCh38
rs577899103 Chr.4: 41745444-41745444 on Build GRCh38
rs957152597 Chr.4: 41745481-41745481 on Build GRCh38
rs547049342 Chr.4: 41745603-41745603 on Build GRCh38
rs1036904128 Chr.4: 41745573-41745573 on Build GRCh38
rs763380864 Chr.4: 41745775-41745777 on Build GRCh38
rs957517040 Chr.4: 41745729-41745729 on Build GRCh38
rs762497253 Chr.4: 41745801-41745801 on Build GRCh38
rs558416040 Chr.4: 41745747-41745747 on Build GRCh38
rs1045005323 Chr.4: 41745831-41745831 on Build GRCh38
rs911357344 Chr.4: 41745772-41745772 on Build GRCh38
rs767873201 Chr.4: 41745823-41745823 on Build GRCh38

View Full Product Details