Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 161349252-161349754 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PARK2
Entrez Gene ID
5071
Gene Name
parkin RBR E3 ubiquitin protein ligase
Gene Aliases
AR-JP, LPRS2, PDJ, PRKN
UniGene
Hs.132954
Gene Chromosome Location
Chr.6: 161347417-162727802 on Build GRCh38
Transcripts
NM_004562.2 NM_013987.2 NM_013988.2 XM_011535863.1 XM_017010908.1 XM_017010909.1

SNP Detail

SNP ID Chromosome Location
rs975312899 Chr.6: 161349632-161349632 on Build GRCh38
rs565359877 Chr.6: 161349470-161349470 on Build GRCh38
rs760233938 Chr.6: 161349446-161349446 on Build GRCh38
rs528792599 Chr.6: 161349424-161349424 on Build GRCh38
rs1047898450 Chr.6: 161349312-161349312 on Build GRCh38
rs902436659 Chr.6: 161349456-161349456 on Build GRCh38
rs112757570 Chr.6: 161349571-161349571 on Build GRCh38
rs965011358 Chr.6: 161349502-161349502 on Build GRCh38
rs764753874 Chr.6: 161349727-161349727 on Build GRCh38
rs943928519 Chr.6: 161349622-161349622 on Build GRCh38
rs776047624 Chr.6: 161349369-161349369 on Build GRCh38
rs71653629 Chr.6: 161349447-161349447 on Build GRCh38
rs912250905 Chr.6: 161349590-161349590 on Build GRCh38
rs931149619 Chr.6: 161349654-161349654 on Build GRCh38
rs950999019 Chr.6: 161349636-161349636 on Build GRCh38
rs942110218 Chr.6: 161349325-161349325 on Build GRCh38
rs940809817 Chr.6: 161349705-161349705 on Build GRCh38
rs1055099691 Chr.6: 161349352-161349352 on Build GRCh38
rs988197174 Chr.6: 161349544-161349544 on Build GRCh38
rs530999210 Chr.6: 161349476-161349476 on Build GRCh38
rs567299724 Chr.6: 161349515-161349515 on Build GRCh38
rs776532449 Chr.6: 161349558-161349561 on Build GRCh38
rs1030964785 Chr.6: 161349542-161349542 on Build GRCh38
rs767963623 Chr.6: 161349287-161349287 on Build GRCh38
rs909255211 Chr.6: 161349318-161349318 on Build GRCh38
rs756738207 Chr.6: 161349694-161349694 on Build GRCh38
rs1032155646 Chr.6: 161349469-161349469 on Build GRCh38
rs74701717 Chr.6: 161349279-161349279 on Build GRCh38
rs1006582328 Chr.6: 161349484-161349484 on Build GRCh38
rs969554917 Chr.6: 161349404-161349404 on Build GRCh38
rs548853930 Chr.6: 161349642-161349642 on Build GRCh38
rs758368939 Chr.6: 161349729-161349729 on Build GRCh38
rs530081303 Chr.6: 161349549-161349549 on Build GRCh38
rs1013865134 Chr.6: 161349284-161349284 on Build GRCh38
rs753440085 Chr.6: 161349573-161349573 on Build GRCh38
rs565924215 Chr.6: 161349651-161349651 on Build GRCh38
rs999729613 Chr.6: 161349466-161349466 on Build GRCh38
rs1048255222 Chr.6: 161349661-161349661 on Build GRCh38
rs886061235 Chr.6: 161349627-161349627 on Build GRCh38
rs942203127 Chr.6: 161349665-161349665 on Build GRCh38
rs188421588 Chr.6: 161349491-161349491 on Build GRCh38
rs895132585 Chr.6: 161349376-161349376 on Build GRCh38

View Full Product Details