Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 161359849-161360364 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PARK2
Entrez Gene ID
5071
Gene Name
parkin RBR E3 ubiquitin protein ligase
Gene Aliases
AR-JP, LPRS2, PDJ, PRKN
UniGene
Hs.132954
Gene Chromosome Location
Chr.6: 161347417-162727802 on Build GRCh38
Transcripts
NM_004562.2 NM_013987.2 NM_013988.2 XM_011535863.1 XM_017010908.1 XM_017010909.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6952784 c.1174A>G p.R392G Chr.6: 161360199-161360199 on Build GRCh38
483667 c.1269A>G p.V423V Chr.6: 161360104-161360104 on Build GRCh38
5667713 c.1245C>A p.T415T Chr.6: 161360128-161360128 on Build GRCh38
6949429 c.1224A>C p.K408N Chr.6: 161360149-161360149 on Build GRCh38
1075794 c.1190C>T p.A397V Chr.6: 161360183-161360183 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765846345 Chr.6: 161360101-161360101 on Build GRCh38
rs760540631 Chr.6: 161360083-161360083 on Build GRCh38
rs1034201351 Chr.6: 161359982-161359982 on Build GRCh38
rs756244498 Chr.6: 161360214-161360214 on Build GRCh38
rs576493932 Chr.6: 161359939-161359939 on Build GRCh38
rs766875584 Chr.6: 161359963-161359963 on Build GRCh38
rs752418689 Chr.6: 161360118-161360118 on Build GRCh38
rs1034293314 Chr.6: 161360315-161360315 on Build GRCh38
rs571092914 Chr.6: 161360181-161360181 on Build GRCh38
rs372141916 Chr.6: 161360179-161360179 on Build GRCh38
rs373028813 Chr.6: 161359985-161359985 on Build GRCh38
rs904536848 Chr.6: 161360186-161360186 on Build GRCh38
rs535800490 Chr.6: 161360253-161360253 on Build GRCh38
rs773142133 Chr.6: 161360172-161360172 on Build GRCh38
rs767878841 Chr.6: 161360202-161360202 on Build GRCh38
rs1048934060 Chr.6: 161360279-161360279 on Build GRCh38
rs952680771 Chr.6: 161359951-161359951 on Build GRCh38
rs1018676251 Chr.6: 161359962-161359962 on Build GRCh38
rs555480305 Chr.6: 161360271-161360271 on Build GRCh38
rs777708182 Chr.6: 161360205-161360205 on Build GRCh38
rs758718482 Chr.6: 161360089-161360090 on Build GRCh38
rs1001601804 Chr.6: 161360290-161360290 on Build GRCh38
rs759137723 Chr.6: 161360108-161360108 on Build GRCh38
rs781138283 Chr.6: 161360043-161360043 on Build GRCh38
rs145068298 Chr.6: 161360316-161360316 on Build GRCh38
rs771931085 Chr.6: 161360171-161360171 on Build GRCh38
rs770987966 Chr.6: 161360040-161360040 on Build GRCh38
rs570053526 Chr.6: 161360212-161360212 on Build GRCh38
rs539917500 Chr.6: 161360187-161360187 on Build GRCh38
rs369693402 Chr.6: 161360046-161360046 on Build GRCh38
rs1027104979 Chr.6: 161359903-161359903 on Build GRCh38
rs531247345 Chr.6: 161360114-161360114 on Build GRCh38
rs562362828 Chr.6: 161360150-161360150 on Build GRCh38
rs540375416 Chr.6: 161359915-161359915 on Build GRCh38
rs780291252 Chr.6: 161360228-161360228 on Build GRCh38
rs151065730 Chr.6: 161359886-161359886 on Build GRCh38
rs371266389 Chr.6: 161360229-161360229 on Build GRCh38
rs58063982 Chr.6: 161359874-161359874 on Build GRCh38
rs746922936 Chr.6: 161360178-161360178 on Build GRCh38
rs370186221 Chr.6: 161360230-161360230 on Build GRCh38
rs888542892 Chr.6: 161360282-161360282 on Build GRCh38
rs757738248 Chr.6: 161360132-161360132 on Build GRCh38
rs778125254 Chr.6: 161360129-161360129 on Build GRCh38
rs376662625 Chr.6: 161360062-161360062 on Build GRCh38
rs920265563 Chr.6: 161360042-161360042 on Build GRCh38
rs773265265 Chr.6: 161360081-161360081 on Build GRCh38
rs960068236 Chr.6: 161360325-161360325 on Build GRCh38
rs55830907 Chr.6: 161360169-161360169 on Build GRCh38
rs753313285 Chr.6: 161360107-161360107 on Build GRCh38
rs113185630 Chr.6: 161359944-161359944 on Build GRCh38
rs754719162 Chr.6: 161359990-161359990 on Build GRCh38
rs371251606 Chr.6: 161359932-161359932 on Build GRCh38
rs868336390 Chr.6: 161360111-161360111 on Build GRCh38
rs140259757 Chr.6: 161359974-161359974 on Build GRCh38
rs951686418 Chr.6: 161360278-161360278 on Build GRCh38
rs886061237 Chr.6: 161360160-161360160 on Build GRCh38
rs994658470 Chr.6: 161359892-161359892 on Build GRCh38
rs766915327 Chr.6: 161360168-161360168 on Build GRCh38
rs751005126 Chr.6: 161360204-161360204 on Build GRCh38
rs762599162 Chr.6: 161360191-161360191 on Build GRCh38
rs1014577040 Chr.6: 161360326-161360326 on Build GRCh38
rs200290839 Chr.6: 161359882-161359882 on Build GRCh38
rs761218733 Chr.6: 161359934-161359934 on Build GRCh38
rs745912659 Chr.6: 161360141-161360141 on Build GRCh38
rs995928189 Chr.6: 161360233-161360233 on Build GRCh38
rs764789423 Chr.6: 161360182-161360182 on Build GRCh38
rs763354380 Chr.6: 161359949-161359949 on Build GRCh38
rs868083234 Chr.6: 161360137-161360137 on Build GRCh38
rs145135117 Chr.6: 161360115-161360115 on Build GRCh38
rs769929840 Chr.6: 161360052-161360052 on Build GRCh38
rs539790024 Chr.6: 161360198-161360198 on Build GRCh38
rs772055546 Chr.6: 161360056-161360056 on Build GRCh38
rs757005515 Chr.6: 161360339-161360339 on Build GRCh38
rs189941210 Chr.6: 161360122-161360122 on Build GRCh38
rs761669123 Chr.6: 161360055-161360055 on Build GRCh38
rs373937983 Chr.6: 161360054-161360054 on Build GRCh38
rs865798516 Chr.6: 161359994-161359994 on Build GRCh38
rs752038413 Chr.6: 161360131-161360131 on Build GRCh38
rs74987477 Chr.6: 161360281-161360281 on Build GRCh38
rs751890444 Chr.6: 161360194-161360194 on Build GRCh38
rs781282224 Chr.6: 161360140-161360140 on Build GRCh38
rs776775740 Chr.6: 161359881-161359881 on Build GRCh38
rs1801334 Chr.6: 161360193-161360193 on Build GRCh38
rs113975640 Chr.6: 161360224-161360224 on Build GRCh38

View Full Product Details