Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 30975968-30976509 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GHRHR
Entrez Gene ID
2692
Gene Name
growth hormone releasing hormone receptor
Gene Aliases
GHRFR, GRFR, IGHD1B
UniGene
Hs.733003
Gene Chromosome Location
Chr.7: 30964021-30979531 on Build GRCh38
Transcripts
NM_000823.3 XM_011515263.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3637969 c.990G>A p.S330S Chr.7: 30976444-30976444 on Build GRCh38
5937299 c.975-7C>T p.? Chr.7: 30976422-30976422 on Build GRCh38
4637178 c.1011C>G p.L337L Chr.7: 30976465-30976465 on Build GRCh38
2149936 c.977G>A p.R326H Chr.7: 30976431-30976431 on Build GRCh38
5791327 c.983C>A p.S328Y Chr.7: 30976437-30976437 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs140322604 Chr.7: 30976216-30976216 on Build GRCh38
rs752122561 Chr.7: 30976443-30976443 on Build GRCh38
rs553323365 Chr.7: 30976152-30976152 on Build GRCh38
rs149182247 Chr.7: 30976461-30976461 on Build GRCh38
rs375789396 Chr.7: 30976425-30976425 on Build GRCh38
rs1057002731 Chr.7: 30975994-30975994 on Build GRCh38
rs887428988 Chr.7: 30976071-30976071 on Build GRCh38
rs538476438 Chr.7: 30976019-30976019 on Build GRCh38
rs202089170 Chr.7: 30976463-30976463 on Build GRCh38
rs750488020 Chr.7: 30976468-30976468 on Build GRCh38
rs372024874 Chr.7: 30976481-30976481 on Build GRCh38
rs529170179 Chr.7: 30976309-30976309 on Build GRCh38
rs779352541 Chr.7: 30976346-30976346 on Build GRCh38
rs117572522 Chr.7: 30976423-30976423 on Build GRCh38
rs990011186 Chr.7: 30976233-30976233 on Build GRCh38
rs4988503 Chr.7: 30976329-30976329 on Build GRCh38
rs1013162125 Chr.7: 30976066-30976066 on Build GRCh38
rs981143509 Chr.7: 30976274-30976274 on Build GRCh38
rs188490880 Chr.7: 30976411-30976411 on Build GRCh38
rs535727568 Chr.7: 30976431-30976431 on Build GRCh38
rs752329366 Chr.7: 30976187-30976187 on Build GRCh38
rs1002879012 Chr.7: 30976164-30976164 on Build GRCh38
rs371934376 Chr.7: 30976378-30976378 on Build GRCh38
rs4988504 Chr.7: 30976403-30976403 on Build GRCh38
rs975868805 Chr.7: 30976202-30976202 on Build GRCh38
rs1038552744 Chr.7: 30976013-30976013 on Build GRCh38
rs762287877 Chr.7: 30976046-30976046 on Build GRCh38
rs121918121 Chr.7: 30976439-30976439 on Build GRCh38
rs1017273353 Chr.7: 30976157-30976157 on Build GRCh38
rs73307743 Chr.7: 30976082-30976082 on Build GRCh38
rs767435657 Chr.7: 30976106-30976106 on Build GRCh38
rs572036013 Chr.7: 30976167-30976167 on Build GRCh38
rs549927188 Chr.7: 30976420-30976420 on Build GRCh38
rs10245096 Chr.7: 30976264-30976264 on Build GRCh38
rs914522016 Chr.7: 30976263-30976263 on Build GRCh38
rs1007818013 Chr.7: 30976350-30976350 on Build GRCh38
rs898010721 Chr.7: 30975999-30975999 on Build GRCh38
rs763314210 Chr.7: 30976430-30976430 on Build GRCh38
rs369089111 Chr.7: 30976444-30976444 on Build GRCh38
rs780426452 Chr.7: 30976480-30976480 on Build GRCh38
rs1002859620 Chr.7: 30976477-30976477 on Build GRCh38
rs527722860 Chr.7: 30976383-30976383 on Build GRCh38
rs36115000 Chr.7: 30976225-30976225 on Build GRCh38
rs576157029 Chr.7: 30976256-30976256 on Build GRCh38
rs756177266 Chr.7: 30976471-30976471 on Build GRCh38
rs939404386 Chr.7: 30976305-30976305 on Build GRCh38
rs556340696 Chr.7: 30976047-30976047 on Build GRCh38
rs552944629 Chr.7: 30976065-30976065 on Build GRCh38
rs774192997 Chr.7: 30976022-30976022 on Build GRCh38
rs754054724 Chr.7: 30976484-30976484 on Build GRCh38
rs1011803516 Chr.7: 30976227-30976227 on Build GRCh38
rs926654684 Chr.7: 30976295-30976295 on Build GRCh38
rs1027933523 Chr.7: 30976024-30976024 on Build GRCh38
rs561875648 Chr.7: 30976306-30976306 on Build GRCh38
rs1049334474 Chr.7: 30976330-30976330 on Build GRCh38
rs954344087 Chr.7: 30976347-30976347 on Build GRCh38
rs749124863 Chr.7: 30976379-30976379 on Build GRCh38
rs919407887 Chr.7: 30976316-30976316 on Build GRCh38

View Full Product Details