Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 66629054-66629566 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCTD7
Entrez Gene ID
154881
Gene Name
potassium channel tetramerization domain containing 7
Gene Aliases
CLN14, EPM3
UniGene
Hs.546627
Gene Chromosome Location
Chr.7: 66628881-66643229 on Build GRCh38
Transcripts
NM_001167961.2 NM_153033.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6391919 c.86A>T p.E29V Chr.7: 66629150-66629150 on Build GRCh38
6408632 c.50C>T p.A17V Chr.7: 66629114-66629114 on Build GRCh38
7314708 c.89_100del12 p.P33_T36delPTAT Chr.7: 66629153-66629164 on Build GRCh38
6988019 c.35G>T p.R12L Chr.7: 66629099-66629099 on Build GRCh38
6343921 c.121G>T p.A41S Chr.7: 66629185-66629185 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs868184670 Chr.7: 66629124-66629124 on Build GRCh38
rs1003950855 Chr.7: 66629324-66629324 on Build GRCh38
rs933703243 Chr.7: 66629470-66629470 on Build GRCh38
rs1052586558 Chr.7: 66629267-66629267 on Build GRCh38
rs934206112 Chr.7: 66629384-66629384 on Build GRCh38
rs975133654 Chr.7: 66629446-66629446 on Build GRCh38
rs561150606 Chr.7: 66629433-66629433 on Build GRCh38
rs984162483 Chr.7: 66629331-66629331 on Build GRCh38
rs886062413 Chr.7: 66629082-66629082 on Build GRCh38
rs549329614 Chr.7: 66629484-66629484 on Build GRCh38
rs753471689 Chr.7: 66629125-66629125 on Build GRCh38
rs934155149 Chr.7: 66629513-66629513 on Build GRCh38
rs757910819 Chr.7: 66629228-66629228 on Build GRCh38
rs587780370 Chr.7: 66629197-66629197 on Build GRCh38
rs978932095 Chr.7: 66629292-66629292 on Build GRCh38
rs933610815 Chr.7: 66629076-66629076 on Build GRCh38
rs149409999 Chr.7: 66629369-66629369 on Build GRCh38
rs763086570 Chr.7: 66629180-66629180 on Build GRCh38
rs761974240 Chr.7: 66629159-66629159 on Build GRCh38
rs915875049 Chr.7: 66629273-66629273 on Build GRCh38
rs766882316 Chr.7: 66629101-66629101 on Build GRCh38
rs953647233 Chr.7: 66629307-66629307 on Build GRCh38
rs757018925 Chr.7: 66629212-66629212 on Build GRCh38
rs962484084 Chr.7: 66629423-66629423 on Build GRCh38
rs778676871 Chr.7: 66629216-66629216 on Build GRCh38
rs1023692565 Chr.7: 66629313-66629313 on Build GRCh38
rs1031987829 Chr.7: 66629252-66629252 on Build GRCh38
rs373655601 Chr.7: 66629177-66629177 on Build GRCh38
rs940166432 Chr.7: 66629358-66629358 on Build GRCh38
rs765260614 Chr.7: 66629096-66629096 on Build GRCh38
rs752044252 Chr.7: 66629192-66629192 on Build GRCh38
rs922774213 Chr.7: 66629382-66629382 on Build GRCh38
rs1007276581 Chr.7: 66629145-66629145 on Build GRCh38
rs998736066 Chr.7: 66629227-66629227 on Build GRCh38
rs544784365 Chr.7: 66629200-66629200 on Build GRCh38
rs893987663 Chr.7: 66629545-66629545 on Build GRCh38
rs1052660198 Chr.7: 66629539-66629539 on Build GRCh38
rs893953846 Chr.7: 66629269-66629269 on Build GRCh38
rs755360166 Chr.7: 66629113-66629113 on Build GRCh38
rs12531758 Chr.7: 66629477-66629477 on Build GRCh38
rs769985127 Chr.7: 66629166-66629166 on Build GRCh38
rs538905437 Chr.7: 66629369-66629370 on Build GRCh38
rs751937729 Chr.7: 66629109-66629109 on Build GRCh38
rs1014094217 Chr.7: 66629469-66629469 on Build GRCh38
rs144762900 Chr.7: 66629373-66629373 on Build GRCh38
rs1022873245 Chr.7: 66629471-66629471 on Build GRCh38
rs984110156 Chr.7: 66629473-66629473 on Build GRCh38
rs549371655 Chr.7: 66629532-66629532 on Build GRCh38
rs778647590 Chr.7: 66629165-66629176 on Build GRCh38
rs754475798 Chr.7: 66629163-66629174 on Build GRCh38
rs970029736 Chr.7: 66629472-66629472 on Build GRCh38
rs926100634 Chr.7: 66629296-66629296 on Build GRCh38
rs888749851 Chr.7: 66629389-66629389 on Build GRCh38
rs890695139 Chr.7: 66629153-66629153 on Build GRCh38
rs773209854 Chr.7: 66629171-66629171 on Build GRCh38
rs1031481633 Chr.7: 66629436-66629436 on Build GRCh38
rs935541837 Chr.7: 66629266-66629266 on Build GRCh38
rs753563583 Chr.7: 66629205-66629205 on Build GRCh38
rs371919994 Chr.7: 66629140-66629140 on Build GRCh38
rs575504075 Chr.7: 66629388-66629388 on Build GRCh38
rs767949338 Chr.7: 66629198-66629198 on Build GRCh38
rs756855275 Chr.7: 66629127-66629127 on Build GRCh38
rs777578179 Chr.7: 66629117-66629117 on Build GRCh38
rs763423181 Chr.7: 66629099-66629099 on Build GRCh38
rs986360420 Chr.7: 66629314-66629314 on Build GRCh38
rs765754234 Chr.7: 66629403-66629403 on Build GRCh38
rs1011009307 Chr.7: 66629271-66629271 on Build GRCh38
rs796052690 Chr.7: 66629114-66629114 on Build GRCh38
rs761888221 Chr.7: 66629094-66629094 on Build GRCh38
rs921008210 Chr.7: 66629447-66629447 on Build GRCh38
rs753399965 Chr.7: 66629153-66629164 on Build GRCh38
rs749965828 Chr.7: 66629218-66629218 on Build GRCh38
rs796052687 Chr.7: 66629204-66629204 on Build GRCh38
rs866407655 Chr.7: 66629396-66629396 on Build GRCh38
rs970042728 Chr.7: 66629323-66629323 on Build GRCh38
rs985702533 Chr.7: 66629519-66629519 on Build GRCh38
rs778513261 Chr.7: 66629130-66629130 on Build GRCh38
rs905195884 Chr.7: 66629317-66629317 on Build GRCh38
rs912120487 Chr.7: 66629337-66629337 on Build GRCh38
rs771944011 Chr.7: 66629137-66629137 on Build GRCh38
rs911513429 Chr.7: 66629543-66629543 on Build GRCh38
rs750951765 Chr.7: 66629098-66629098 on Build GRCh38
rs544446477 Chr.7: 66629395-66629395 on Build GRCh38
rs745360140 Chr.7: 66629132-66629132 on Build GRCh38
rs961457138 Chr.7: 66629502-66629502 on Build GRCh38
rs776871737 Chr.7: 66629157-66629157 on Build GRCh38
rs766856368 Chr.7: 66629185-66629185 on Build GRCh38
rs559492597 Chr.7: 66629503-66629503 on Build GRCh38
rs768374078 Chr.7: 66629150-66629150 on Build GRCh38
rs912169541 Chr.7: 66629483-66629483 on Build GRCh38
rs910008109 Chr.7: 66629344-66629344 on Build GRCh38
rs1016743099 Chr.7: 66629411-66629411 on Build GRCh38

View Full Product Details