Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 66633186-66633685 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCTD7
Entrez Gene ID
154881
Gene Name
potassium channel tetramerization domain containing 7
Gene Aliases
CLN14, EPM3
UniGene
Hs.546627
Gene Chromosome Location
Chr.7: 66628881-66643229 on Build GRCh38
Transcripts
NM_001167961.2 NM_153033.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3640112 c.268G>A p.D90N Chr.7: 66633398-66633398 on Build GRCh38
1091327 c.162C>T p.P54P Chr.7: 66633292-66633292 on Build GRCh38
5716570 c.249G>T p.G83G Chr.7: 66633379-66633379 on Build GRCh38
7373311 c.292G>A p.D98N Chr.7: 66633422-66633422 on Build GRCh38
5480700 c.267G>A p.T89T Chr.7: 66633397-66633397 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746002015 Chr.7: 66633400-66633400 on Build GRCh38
rs183146094 Chr.7: 66633650-66633650 on Build GRCh38
rs3764902 Chr.7: 66633595-66633595 on Build GRCh38
rs200569899 Chr.7: 66633356-66633356 on Build GRCh38
rs765917850 Chr.7: 66633370-66633370 on Build GRCh38
rs368755155 Chr.7: 66633271-66633271 on Build GRCh38
rs770973872 Chr.7: 66633270-66633270 on Build GRCh38
rs776037212 Chr.7: 66633290-66633290 on Build GRCh38
rs752248447 Chr.7: 66633302-66633302 on Build GRCh38
rs755849045 Chr.7: 66633325-66633325 on Build GRCh38
rs543103689 Chr.7: 66633256-66633256 on Build GRCh38
rs188483031 Chr.7: 66633387-66633387 on Build GRCh38
rs867319899 Chr.7: 66633338-66633338 on Build GRCh38
rs200575329 Chr.7: 66633381-66633381 on Build GRCh38
rs3764903 Chr.7: 66633495-66633495 on Build GRCh38
rs745373183 Chr.7: 66633549-66633549 on Build GRCh38
rs387907260 Chr.7: 66633410-66633410 on Build GRCh38
rs797005416 Chr.7: 66633593-66633594 on Build GRCh38
rs923747930 Chr.7: 66633479-66633479 on Build GRCh38
rs1041847363 Chr.7: 66633621-66633621 on Build GRCh38
rs138642014 Chr.7: 66633357-66633357 on Build GRCh38
rs919262794 Chr.7: 66633611-66633611 on Build GRCh38
rs932041772 Chr.7: 66633556-66633556 on Build GRCh38
rs776125299 Chr.7: 66633351-66633351 on Build GRCh38
rs202167686 Chr.7: 66633369-66633369 on Build GRCh38
rs930526100 Chr.7: 66633259-66633259 on Build GRCh38
rs374702986 Chr.7: 66633254-66633254 on Build GRCh38
rs750033880 Chr.7: 66633302-66633302 on Build GRCh38
rs914692626 Chr.7: 66633611-66633611 on Build GRCh38
rs267607199 Chr.7: 66633425-66633425 on Build GRCh38
rs375586745 Chr.7: 66633396-66633396 on Build GRCh38
rs761117000 Chr.7: 66633292-66633292 on Build GRCh38
rs368018261 Chr.7: 66633311-66633311 on Build GRCh38
rs762081281 Chr.7: 66633453-66633453 on Build GRCh38
rs985251877 Chr.7: 66633394-66633394 on Build GRCh38
rs951952763 Chr.7: 66633405-66633405 on Build GRCh38
rs139585796 Chr.7: 66633403-66633403 on Build GRCh38
rs529063727 Chr.7: 66633312-66633312 on Build GRCh38
rs200176641 Chr.7: 66633594-66633595 on Build GRCh38
rs976005236 Chr.7: 66633527-66633527 on Build GRCh38
rs780710316 Chr.7: 66633404-66633404 on Build GRCh38
rs371824397 Chr.7: 66633298-66633298 on Build GRCh38
rs202174213 Chr.7: 66633592-66633593 on Build GRCh38
rs752567923 Chr.7: 66633385-66633385 on Build GRCh38
rs368651767 Chr.7: 66633593-66633594 on Build GRCh38
rs374377118 Chr.7: 66633566-66633566 on Build GRCh38
rs770934416 Chr.7: 66633343-66633343 on Build GRCh38
rs781014688 Chr.7: 66633242-66633242 on Build GRCh38
rs772773351 Chr.7: 66633347-66633347 on Build GRCh38
rs373509859 Chr.7: 66633408-66633408 on Build GRCh38
rs149255570 Chr.7: 66633386-66633386 on Build GRCh38
rs150589535 Chr.7: 66633355-66633355 on Build GRCh38
rs752649838 Chr.7: 66633473-66633473 on Build GRCh38
rs536010561 Chr.7: 66633618-66633618 on Build GRCh38
rs369305390 Chr.7: 66633468-66633468 on Build GRCh38
rs749483829 Chr.7: 66633339-66633339 on Build GRCh38
rs751140440 Chr.7: 66633374-66633374 on Build GRCh38
rs376524066 Chr.7: 66633301-66633301 on Build GRCh38
rs555702878 Chr.7: 66633634-66633634 on Build GRCh38
rs768463613 Chr.7: 66633241-66633241 on Build GRCh38
rs769023482 Chr.7: 66633421-66633421 on Build GRCh38
rs779862565 Chr.7: 66633238-66633238 on Build GRCh38
rs770677527 Chr.7: 66633455-66633455 on Build GRCh38
rs142526573 Chr.7: 66633322-66633322 on Build GRCh38
rs760713984 Chr.7: 66633476-66633476 on Build GRCh38
rs35686816 Chr.7: 66633445-66633446 on Build GRCh38
rs763903390 Chr.7: 66633494-66633494 on Build GRCh38
rs769204848 Chr.7: 66633345-66633345 on Build GRCh38
rs3764904 Chr.7: 66633397-66633397 on Build GRCh38
rs140163210 Chr.7: 66633642-66633642 on Build GRCh38
rs757879423 Chr.7: 66633308-66633308 on Build GRCh38
rs745910052 Chr.7: 66633346-66633346 on Build GRCh38
rs1053905845 Chr.7: 66633507-66633507 on Build GRCh38
rs903362733 Chr.7: 66633623-66633626 on Build GRCh38
rs754476100 Chr.7: 66633380-66633380 on Build GRCh38
rs777859099 Chr.7: 66633334-66633334 on Build GRCh38
rs200321023 Chr.7: 66633323-66633323 on Build GRCh38
rs1003043263 Chr.7: 66633625-66633628 on Build GRCh38
rs894847620 Chr.7: 66633509-66633509 on Build GRCh38
rs947473989 Chr.7: 66633615-66633615 on Build GRCh38
rs559020294 Chr.7: 66633275-66633275 on Build GRCh38
rs984651812 Chr.7: 66633411-66633411 on Build GRCh38
rs749343374 Chr.7: 66633260-66633260 on Build GRCh38
rs200130485 Chr.7: 66633594-66633594 on Build GRCh38
rs752696472 Chr.7: 66633449-66633449 on Build GRCh38
rs749010969 Chr.7: 66633388-66633388 on Build GRCh38
rs201296399 Chr.7: 66633320-66633320 on Build GRCh38
rs747910548 Chr.7: 66633248-66633248 on Build GRCh38
rs768112652 Chr.7: 66633280-66633280 on Build GRCh38
rs766971101 Chr.7: 66633469-66633469 on Build GRCh38
rs1011937355 Chr.7: 66633659-66633659 on Build GRCh38
rs556693792 Chr.7: 66633463-66633463 on Build GRCh38
rs3764905 Chr.7: 66633239-66633239 on Build GRCh38

View Full Product Details