Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 66639167-66639679 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCTD7
Entrez Gene ID
154881
Gene Name
potassium channel tetramerization domain containing 7
Gene Aliases
CLN14, EPM3
UniGene
Hs.546627
Gene Chromosome Location
Chr.7: 66628881-66643229 on Build GRCh38
Transcripts
NM_001167961.2 NM_153033.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
254756 c.847G>C p.E283Q Chr.7: 66639209-66639209 on Build GRCh38
80044 c.816G>C p.V272V Chr.7: 66639178-66639178 on Build GRCh38
3640118 c.820C>T p.H274Y Chr.7: 66639182-66639182 on Build GRCh38
1292534 c.838C>T p.P280S Chr.7: 66639200-66639200 on Build GRCh38
3640119 c.839C>T p.P280L Chr.7: 66639201-66639201 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs951146954 Chr.7: 66639537-66639537 on Build GRCh38
rs922192298 Chr.7: 66639647-66639647 on Build GRCh38
rs748183090 Chr.7: 66639225-66639225 on Build GRCh38
rs1023493281 Chr.7: 66639454-66639454 on Build GRCh38
rs932179746 Chr.7: 66639377-66639377 on Build GRCh38
rs940752551 Chr.7: 66639349-66639349 on Build GRCh38
rs758401521 Chr.7: 66639284-66639284 on Build GRCh38
rs907869743 Chr.7: 66639346-66639346 on Build GRCh38
rs756747620 Chr.7: 66639256-66639256 on Build GRCh38
rs953482106 Chr.7: 66639546-66639546 on Build GRCh38
rs773953605 Chr.7: 66639236-66639236 on Build GRCh38
rs367591875 Chr.7: 66639223-66639223 on Build GRCh38
rs774927044 Chr.7: 66639242-66639242 on Build GRCh38
rs116630203 Chr.7: 66639241-66639241 on Build GRCh38
rs923327947 Chr.7: 66639544-66639546 on Build GRCh38
rs749877863 Chr.7: 66639266-66639266 on Build GRCh38
rs376744529 Chr.7: 66639595-66639595 on Build GRCh38
rs1014919686 Chr.7: 66639500-66639500 on Build GRCh38
rs763849444 Chr.7: 66639248-66639248 on Build GRCh38
rs960570960 Chr.7: 66639222-66639222 on Build GRCh38
rs113446781 Chr.7: 66639383-66639383 on Build GRCh38
rs974095274 Chr.7: 66639624-66639624 on Build GRCh38
rs879662632 Chr.7: 66639607-66639607 on Build GRCh38
rs145063728 Chr.7: 66639197-66639197 on Build GRCh38
rs1042399199 Chr.7: 66639610-66639610 on Build GRCh38
rs962369833 Chr.7: 66639620-66639620 on Build GRCh38
rs951958149 Chr.7: 66639263-66639263 on Build GRCh38
rs969666860 Chr.7: 66639476-66639476 on Build GRCh38
rs923614735 Chr.7: 66639652-66639652 on Build GRCh38
rs986221954 Chr.7: 66639556-66639556 on Build GRCh38
rs541759714 Chr.7: 66639382-66639382 on Build GRCh38
rs923444735 Chr.7: 66639365-66639365 on Build GRCh38
rs746774522 Chr.7: 66639196-66639196 on Build GRCh38
rs976607982 Chr.7: 66639351-66639351 on Build GRCh38
rs563887098 Chr.7: 66639634-66639634 on Build GRCh38
rs922098847 Chr.7: 66639336-66639336 on Build GRCh38
rs768574038 Chr.7: 66639203-66639203 on Build GRCh38
rs149123496 Chr.7: 66639334-66639334 on Build GRCh38
rs1052877177 Chr.7: 66639431-66639431 on Build GRCh38
rs151151235 Chr.7: 66639453-66639453 on Build GRCh38
rs77341088 Chr.7: 66639448-66639448 on Build GRCh38
rs1003311931 Chr.7: 66639477-66639477 on Build GRCh38
rs947639717 Chr.7: 66639391-66639391 on Build GRCh38
rs779871558 Chr.7: 66639194-66639194 on Build GRCh38
rs1034760479 Chr.7: 66639529-66639529 on Build GRCh38
rs776525362 Chr.7: 66639438-66639438 on Build GRCh38
rs759961871 Chr.7: 66639247-66639247 on Build GRCh38
rs375872253 Chr.7: 66639240-66639240 on Build GRCh38
rs1035207417 Chr.7: 66639443-66639443 on Build GRCh38
rs753471697 Chr.7: 66639254-66639254 on Build GRCh38
rs985121432 Chr.7: 66639469-66639469 on Build GRCh38
rs772819046 Chr.7: 66639519-66639519 on Build GRCh38
rs764754359 Chr.7: 66639260-66639260 on Build GRCh38
rs886062414 Chr.7: 66639287-66639287 on Build GRCh38
rs890525029 Chr.7: 66639392-66639392 on Build GRCh38
rs932169084 Chr.7: 66639653-66639653 on Build GRCh38
rs893768954 Chr.7: 66639432-66639432 on Build GRCh38
rs769596185 Chr.7: 66639506-66639506 on Build GRCh38
rs909540261 Chr.7: 66639587-66639587 on Build GRCh38
rs768924233 Chr.7: 66639198-66639198 on Build GRCh38
rs999586476 Chr.7: 66639415-66639415 on Build GRCh38
rs914747817 Chr.7: 66639573-66639573 on Build GRCh38
rs540871291 Chr.7: 66639586-66639586 on Build GRCh38
rs776777218 Chr.7: 66639214-66639214 on Build GRCh38
rs1037925062 Chr.7: 66639295-66639295 on Build GRCh38

View Full Product Details