Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 66642286-66642782 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCTD7
Entrez Gene ID
154881
Gene Name
potassium channel tetramerization domain containing 7
Gene Aliases
CLN14, EPM3
UniGene
Hs.546627
Gene Chromosome Location
Chr.7: 66628881-66643229 on Build GRCh38
Transcripts
NM_001167961.2 NM_153033.4

SNP Detail

SNP ID Chromosome Location
rs942950804 Chr.7: 66642380-66642380 on Build GRCh38
rs756308299 Chr.7: 66642355-66642355 on Build GRCh38
rs962037284 Chr.7: 66642683-66642683 on Build GRCh38
rs1034118225 Chr.7: 66642310-66642310 on Build GRCh38
rs921599597 Chr.7: 66642387-66642387 on Build GRCh38
rs951434393 Chr.7: 66642638-66642638 on Build GRCh38
rs992496804 Chr.7: 66642324-66642324 on Build GRCh38
rs1048604873 Chr.7: 66642486-66642486 on Build GRCh38
rs567219674 Chr.7: 66642588-66642588 on Build GRCh38
rs891498687 Chr.7: 66642449-66642449 on Build GRCh38
rs931500831 Chr.7: 66642484-66642484 on Build GRCh38
rs1039953803 Chr.7: 66642386-66642386 on Build GRCh38
rs984331973 Chr.7: 66642738-66642738 on Build GRCh38
rs1024005661 Chr.7: 66642676-66642676 on Build GRCh38
rs763823588 Chr.7: 66642469-66642469 on Build GRCh38
rs1042055931 Chr.7: 66642600-66642600 on Build GRCh38
rs894277876 Chr.7: 66642650-66642650 on Build GRCh38
rs911367720 Chr.7: 66642518-66642518 on Build GRCh38
rs1027702204 Chr.7: 66642689-66642689 on Build GRCh38
rs973433847 Chr.7: 66642466-66642466 on Build GRCh38
rs986884390 Chr.7: 66642644-66642644 on Build GRCh38
rs1013992683 Chr.7: 66642675-66642675 on Build GRCh38
rs916921596 Chr.7: 66642346-66642346 on Build GRCh38
rs964207551 Chr.7: 66642684-66642684 on Build GRCh38
rs867326497 Chr.7: 66642315-66642315 on Build GRCh38
rs545267526 Chr.7: 66642752-66642752 on Build GRCh38
rs904452164 Chr.7: 66642609-66642609 on Build GRCh38
rs1009996181 Chr.7: 66642458-66642458 on Build GRCh38
rs573049844 Chr.7: 66642740-66642740 on Build GRCh38
rs962674356 Chr.7: 66642508-66642508 on Build GRCh38
rs144828027 Chr.7: 66642470-66642470 on Build GRCh38
rs571947982 Chr.7: 66642335-66642335 on Build GRCh38
rs553134685 Chr.7: 66642687-66642687 on Build GRCh38
rs925503205 Chr.7: 66642705-66642709 on Build GRCh38
rs1054146651 Chr.7: 66642445-66642445 on Build GRCh38
rs953008144 Chr.7: 66642696-66642696 on Build GRCh38
rs778804133 Chr.7: 66642631-66642631 on Build GRCh38
rs901211446 Chr.7: 66642563-66642563 on Build GRCh38
rs942772041 Chr.7: 66642559-66642559 on Build GRCh38
rs148095742 Chr.7: 66642657-66642657 on Build GRCh38
rs935637314 Chr.7: 66642407-66642407 on Build GRCh38
rs866413605 Chr.7: 66642586-66642586 on Build GRCh38
rs1025648532 Chr.7: 66642602-66642602 on Build GRCh38
rs757187145 Chr.7: 66642562-66642562 on Build GRCh38
rs570351540 Chr.7: 66642573-66642573 on Build GRCh38
rs961727321 Chr.7: 66642409-66642409 on Build GRCh38
rs552278503 Chr.7: 66642448-66642448 on Build GRCh38
rs867199438 Chr.7: 66642724-66642724 on Build GRCh38
rs1000046524 Chr.7: 66642615-66642615 on Build GRCh38
rs907076403 Chr.7: 66642468-66642468 on Build GRCh38

View Full Product Details