Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100629186-100629592 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TFR2
Entrez Gene ID
7036
Gene Name
transferrin receptor 2
Gene Aliases
HFE3, TFRC2
UniGene
Hs.544932
Gene Chromosome Location
Chr.7: 100620416-100642780 on Build GRCh38
Transcripts
NM_001206855.1 NM_003227.3 XM_005250553.4 XM_017012573.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5497143 c.1333G>C p.V445L Chr.7: 100629310-100629310 on Build GRCh38
4903398 c.1273C>T p.H425Y Chr.7: 100629370-100629370 on Build GRCh38
5492271 c.1340C>T p.T447M Chr.7: 100629303-100629303 on Build GRCh38
247799 c.1298G>T p.R433M Chr.7: 100629345-100629345 on Build GRCh38
7393640 c.1364G>A p.R455Q Chr.7: 100629279-100629279 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747965768 Chr.7: 100629356-100629356 on Build GRCh38
rs746034002 Chr.7: 100629368-100629368 on Build GRCh38
rs775508532 Chr.7: 100629406-100629406 on Build GRCh38
rs774991526 Chr.7: 100629299-100629299 on Build GRCh38
rs776917443 Chr.7: 100629416-100629416 on Build GRCh38
rs527570302 Chr.7: 100629427-100629427 on Build GRCh38
rs777130912 Chr.7: 100629221-100629226 on Build GRCh38
rs41303498 Chr.7: 100629428-100629428 on Build GRCh38
rs762216388 Chr.7: 100629364-100629364 on Build GRCh38
rs775827265 Chr.7: 100629380-100629380 on Build GRCh38
rs542021739 Chr.7: 100629213-100629213 on Build GRCh38
rs532187374 Chr.7: 100629302-100629302 on Build GRCh38
rs75009042 Chr.7: 100629547-100629547 on Build GRCh38
rs368298121 Chr.7: 100629314-100629314 on Build GRCh38
rs148855326 Chr.7: 100629272-100629272 on Build GRCh38
rs761211765 Chr.7: 100629408-100629408 on Build GRCh38
rs570000600 Chr.7: 100629468-100629468 on Build GRCh38
rs780133884 Chr.7: 100629254-100629254 on Build GRCh38
rs151173921 Chr.7: 100629310-100629310 on Build GRCh38
rs773399865 Chr.7: 100629552-100629552 on Build GRCh38
rs80338884 Chr.7: 100629313-100629313 on Build GRCh38
rs1039701799 Chr.7: 100629521-100629521 on Build GRCh38
rs772076371 Chr.7: 100629293-100629293 on Build GRCh38
rs535869195 Chr.7: 100629365-100629365 on Build GRCh38
rs960761563 Chr.7: 100629413-100629413 on Build GRCh38
rs41303501 Chr.7: 100629279-100629279 on Build GRCh38
rs887907860 Chr.7: 100629343-100629343 on Build GRCh38
rs780902940 Chr.7: 100629355-100629355 on Build GRCh38
rs267601182 Chr.7: 100629349-100629349 on Build GRCh38
rs199725911 Chr.7: 100629282-100629282 on Build GRCh38
rs757985211 Chr.7: 100629321-100629321 on Build GRCh38
rs1020333883 Chr.7: 100629539-100629539 on Build GRCh38
rs770380557 Chr.7: 100629211-100629212 on Build GRCh38
rs1040654031 Chr.7: 100629292-100629292 on Build GRCh38
rs759974547 Chr.7: 100629222-100629222 on Build GRCh38
rs749498767 Chr.7: 100629280-100629280 on Build GRCh38
rs368929184 Chr.7: 100629412-100629412 on Build GRCh38
rs768843272 Chr.7: 100629298-100629298 on Build GRCh38
rs763348482 Chr.7: 100629406-100629407 on Build GRCh38
rs556009276 Chr.7: 100629426-100629426 on Build GRCh38
rs770391274 Chr.7: 100629221-100629222 on Build GRCh38
rs541478502 Chr.7: 100629520-100629520 on Build GRCh38
rs1020301245 Chr.7: 100629280-100629281 on Build GRCh38
rs751562784 Chr.7: 100629344-100629344 on Build GRCh38
rs774355364 Chr.7: 100629283-100629283 on Build GRCh38
rs1052819097 Chr.7: 100629270-100629270 on Build GRCh38
rs372014293 Chr.7: 100629338-100629338 on Build GRCh38
rs761420974 Chr.7: 100629307-100629307 on Build GRCh38
rs200751287 Chr.7: 100629262-100629262 on Build GRCh38
rs772164296 Chr.7: 100629398-100629398 on Build GRCh38
rs559059469 Chr.7: 100629484-100629484 on Build GRCh38
rs1013990276 Chr.7: 100629448-100629448 on Build GRCh38
rs577373172 Chr.7: 100629510-100629510 on Build GRCh38
rs147982847 Chr.7: 100629286-100629286 on Build GRCh38
rs979923571 Chr.7: 100629209-100629209 on Build GRCh38
rs35416331 Chr.7: 100629383-100629384 on Build GRCh38
rs753456186 Chr.7: 100629210-100629210 on Build GRCh38
rs537393233 Chr.7: 100629469-100629469 on Build GRCh38
rs901198924 Chr.7: 100629532-100629532 on Build GRCh38
rs77802285 Chr.7: 100629538-100629538 on Build GRCh38
rs754922209 Chr.7: 100629354-100629354 on Build GRCh38
rs7457868 Chr.7: 100629565-100629565 on Build GRCh38
rs971821768 Chr.7: 100629447-100629450 on Build GRCh38
rs775080669 Chr.7: 100629234-100629234 on Build GRCh38
rs769402280 Chr.7: 100629223-100629223 on Build GRCh38
rs758466203 Chr.7: 100629218-100629218 on Build GRCh38
rs754200714 Chr.7: 100629303-100629303 on Build GRCh38
rs756893916 Chr.7: 100629212-100629212 on Build GRCh38

View Full Product Details