Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 202940893-202941429 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADIPOR1
Entrez Gene ID
51094
Gene Name
adiponectin receptor 1
Gene Aliases
ACDCR1, CGI-45, CGI45, PAQR1, TESBP1A
UniGene
Hs.5298
Gene Chromosome Location
Chr.1: 202940825-202958572 on Build GRCh38
Transcripts
NM_001290553.1 NM_001290557.1 NM_001290629.1 NM_015999.5 XM_017001407.1

SNP Detail

SNP ID Chromosome Location
rs370317002 Chr.1: 202941264-202941264 on Build GRCh38
rs913124906 Chr.1: 202941276-202941276 on Build GRCh38
rs747941880 Chr.1: 202940949-202940949 on Build GRCh38
rs979837299 Chr.1: 202941311-202941311 on Build GRCh38
rs1055229618 Chr.1: 202941119-202941119 on Build GRCh38
rs759008232 Chr.1: 202941401-202941401 on Build GRCh38
rs551634906 Chr.1: 202940948-202940948 on Build GRCh38
rs181259491 Chr.1: 202941329-202941329 on Build GRCh38
rs923060558 Chr.1: 202940924-202940924 on Build GRCh38
rs146219634 Chr.1: 202941203-202941203 on Build GRCh38
rs988749266 Chr.1: 202941271-202941271 on Build GRCh38
rs8167 Chr.1: 202941059-202941059 on Build GRCh38
rs995891967 Chr.1: 202941237-202941237 on Build GRCh38
rs1055113305 Chr.1: 202941338-202941338 on Build GRCh38
rs80165154 Chr.1: 202940993-202940993 on Build GRCh38
rs949542709 Chr.1: 202941374-202941374 on Build GRCh38
rs878957482 Chr.1: 202941093-202941093 on Build GRCh38
rs938219144 Chr.1: 202941117-202941117 on Build GRCh38
rs773862347 Chr.1: 202941031-202941031 on Build GRCh38
rs10581 Chr.1: 202941190-202941190 on Build GRCh38
rs11547945 Chr.1: 202940981-202940981 on Build GRCh38
rs913920560 Chr.1: 202940981-202940982 on Build GRCh38
rs954504508 Chr.1: 202941268-202941268 on Build GRCh38
rs879234012 Chr.1: 202940982-202940982 on Build GRCh38
rs1048010405 Chr.1: 202941400-202941400 on Build GRCh38
rs776141978 Chr.1: 202941378-202941378 on Build GRCh38
rs533494322 Chr.1: 202941037-202941037 on Build GRCh38
rs117755738 Chr.1: 202941355-202941355 on Build GRCh38
rs113890019 Chr.1: 202941050-202941050 on Build GRCh38
rs1043268 Chr.1: 202941138-202941138 on Build GRCh38
rs1030182297 Chr.1: 202941253-202941253 on Build GRCh38
rs988698175 Chr.1: 202940980-202940980 on Build GRCh38
rs1043280 Chr.1: 202941100-202941100 on Build GRCh38
rs112936913 Chr.1: 202940932-202940932 on Build GRCh38
rs964821090 Chr.1: 202941216-202941216 on Build GRCh38
rs918111374 Chr.1: 202941339-202941339 on Build GRCh38
rs896631477 Chr.1: 202941120-202941120 on Build GRCh38
rs886135045 Chr.1: 202941200-202941200 on Build GRCh38
rs746970526 Chr.1: 202941205-202941205 on Build GRCh38
rs548140409 Chr.1: 202941318-202941318 on Build GRCh38
rs546508410 Chr.1: 202941175-202941175 on Build GRCh38
rs114825641 Chr.1: 202941328-202941328 on Build GRCh38
rs886688881 Chr.1: 202941405-202941405 on Build GRCh38
rs1013811804 Chr.1: 202941128-202941128 on Build GRCh38
rs966767762 Chr.1: 202941297-202941297 on Build GRCh38
rs1043843230 Chr.1: 202940990-202940990 on Build GRCh38

View Full Product Details