Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 49192814-49193363 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOLH1
Entrez Gene ID
2346
Gene Name
folate hydrolase (prostate-specific membrane antigen) 1
Gene Aliases
FGCP, FOLH, GCP2, GCPII, NAALAD1, NAALAdase, PSM, PSMA, mGCP
UniGene
Hs.645352
Gene Chromosome Location
Chr.11: 49145094-49208706 on Build GRCh38
Transcripts
NM_001014986.1 NM_001193471.1 NM_001193472.1 NM_001193473.1 NM_004476.1 XM_011519958.2 XM_017017432.1 XM_017017433.1 XM_017017434.1 XM_017017435.1 XM_017017436.1 XM_017017437.1 XM_017017438.1 XM_017017439.1 XM_017017440.1 XM_017017441.1 XM_017017442.1 XM_017017443.1 XM_017017444.1 XM_017017445.1 XM_017017446.1 XM_017017447.1 XM_017017448.1 XM_017017449.1 XM_017017450.1 XM_017017451.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
927690 c.458A>G p.N153S Chr.11: 49192848-49192848 on Build GRCh38
4192692 c.472G>T p.V158L Chr.11: 49192834-49192834 on Build GRCh38
161123 c.483C>G p.F161L Chr.11: 49192823-49192823 on Build GRCh38
687835 c.487G>T p.A163S Chr.11: 49192819-49192819 on Build GRCh38
6426878 c.430T>C p.F144L Chr.11: 49192876-49192876 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770146374 Chr.11: 49192927-49192927 on Build GRCh38
rs867685160 Chr.11: 49193275-49193275 on Build GRCh38
rs541754555 Chr.11: 49193050-49193050 on Build GRCh38
rs561688059 Chr.11: 49193052-49193052 on Build GRCh38
rs749082249 Chr.11: 49192940-49192940 on Build GRCh38
rs137879711 Chr.11: 49192842-49192842 on Build GRCh38
rs539392299 Chr.11: 49192980-49192980 on Build GRCh38
rs576023201 Chr.11: 49193028-49193028 on Build GRCh38
rs752256226 Chr.11: 49192865-49192865 on Build GRCh38
rs371189186 Chr.11: 49192863-49192863 on Build GRCh38
rs1018536291 Chr.11: 49193035-49193035 on Build GRCh38
rs752417704 Chr.11: 49193116-49193116 on Build GRCh38
rs765524382 Chr.11: 49192881-49192881 on Build GRCh38
rs907643862 Chr.11: 49193125-49193125 on Build GRCh38
rs764234371 Chr.11: 49193200-49193200 on Build GRCh38
rs369560869 Chr.11: 49193293-49193293 on Build GRCh38
rs1045608853 Chr.11: 49192917-49192917 on Build GRCh38
rs780410301 Chr.11: 49192892-49192892 on Build GRCh38
rs1054470751 Chr.11: 49192958-49192958 on Build GRCh38
rs4315497 Chr.11: 49192947-49192947 on Build GRCh38
rs893173087 Chr.11: 49192959-49192959 on Build GRCh38
rs937264713 Chr.11: 49193175-49193175 on Build GRCh38
rs770810342 Chr.11: 49192943-49192943 on Build GRCh38
rs781363048 Chr.11: 49192913-49192913 on Build GRCh38
rs995808511 Chr.11: 49193068-49193068 on Build GRCh38
rs1008393635 Chr.11: 49193337-49193337 on Build GRCh38
rs891286094 Chr.11: 49192993-49192993 on Build GRCh38
rs1027324193 Chr.11: 49193088-49193088 on Build GRCh38
rs571867286 Chr.11: 49193074-49193074 on Build GRCh38
rs1054843169 Chr.11: 49193179-49193179 on Build GRCh38
rs1041786575 Chr.11: 49193297-49193297 on Build GRCh38
rs767098710 Chr.11: 49192859-49192859 on Build GRCh38
rs981278658 Chr.11: 49193138-49193138 on Build GRCh38
rs778208672 Chr.11: 49192938-49192938 on Build GRCh38
rs1008424584 Chr.11: 49193013-49193013 on Build GRCh38
rs144409953 Chr.11: 49192887-49192887 on Build GRCh38
rs758949926 Chr.11: 49192855-49192855 on Build GRCh38
rs199532174 Chr.11: 49192883-49192883 on Build GRCh38
rs779357481 Chr.11: 49192905-49192905 on Build GRCh38
rs951862517 Chr.11: 49193092-49193092 on Build GRCh38
rs770988853 Chr.11: 49192846-49192846 on Build GRCh38
rs375439459 Chr.11: 49192886-49192886 on Build GRCh38
rs752154961 Chr.11: 49192895-49192895 on Build GRCh38
rs891256359 Chr.11: 49193320-49193320 on Build GRCh38
rs760347053 Chr.11: 49192879-49192879 on Build GRCh38
rs748441657 Chr.11: 49192840-49192840 on Build GRCh38
rs34540261 Chr.11: 49192861-49192862 on Build GRCh38
rs377754089 Chr.11: 49192841-49192841 on Build GRCh38
rs577017467 Chr.11: 49192977-49192977 on Build GRCh38
rs754980946 Chr.11: 49192899-49192899 on Build GRCh38
rs748331163 Chr.11: 49192918-49192918 on Build GRCh38
rs143422020 Chr.11: 49193016-49193016 on Build GRCh38
rs971184207 Chr.11: 49193131-49193131 on Build GRCh38
rs534437382 Chr.11: 49192853-49192853 on Build GRCh38
rs1010341618 Chr.11: 49192959-49192962 on Build GRCh38

View Full Product Details