Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 68060978-68061482 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUS2
Entrez Gene ID
54920
Gene Name
dihydrouridine synthase 2
Gene Aliases
DUS2L, SMM1, URLC8
UniGene
Hs.534460
Gene Chromosome Location
Chr.16: 68022944-68079323 on Build GRCh38
Transcripts
NM_001271762.1 NM_001271763.1 NM_017803.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6259703 c.374G>T p.G125V Chr.16: 68061070-68061070 on Build GRCh38
6195218 c.412G>C p.E138Q Chr.16: 68061108-68061108 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs898179687 Chr.16: 68061379-68061379 on Build GRCh38
rs888437785 Chr.16: 68061385-68061385 on Build GRCh38
rs775884124 Chr.16: 68061164-68061164 on Build GRCh38
rs374313855 Chr.16: 68061459-68061459 on Build GRCh38
rs777081000 Chr.16: 68061176-68061176 on Build GRCh38
rs373281928 Chr.16: 68061109-68061109 on Build GRCh38
rs1037484367 Chr.16: 68061390-68061390 on Build GRCh38
rs765562701 Chr.16: 68061047-68061047 on Build GRCh38
rs747613825 Chr.16: 68061127-68061127 on Build GRCh38
rs753480436 Chr.16: 68061085-68061085 on Build GRCh38
rs936469773 Chr.16: 68061247-68061247 on Build GRCh38
rs962182526 Chr.16: 68061151-68061151 on Build GRCh38
rs771380418 Chr.16: 68061016-68061016 on Build GRCh38
rs767315952 Chr.16: 68061062-68061062 on Build GRCh38
rs561762847 Chr.16: 68061196-68061196 on Build GRCh38
rs752768723 Chr.16: 68061048-68061048 on Build GRCh38
rs774825464 Chr.16: 68061406-68061406 on Build GRCh38
rs564043596 Chr.16: 68061171-68061171 on Build GRCh38
rs754554535 Chr.16: 68061090-68061090 on Build GRCh38
rs775882715 Chr.16: 68061335-68061335 on Build GRCh38
rs372151693 Chr.16: 68061178-68061178 on Build GRCh38
rs753703445 Chr.16: 68061022-68061023 on Build GRCh38
rs914073740 Chr.16: 68061291-68061291 on Build GRCh38
rs142644508 Chr.16: 68061126-68061126 on Build GRCh38
rs778991652 Chr.16: 68061001-68061001 on Build GRCh38
rs369465133 Chr.16: 68061123-68061123 on Build GRCh38
rs759992690 Chr.16: 68061141-68061141 on Build GRCh38
rs953323156 Chr.16: 68061445-68061445 on Build GRCh38
rs1003919992 Chr.16: 68061041-68061041 on Build GRCh38
rs776708464 Chr.16: 68061028-68061028 on Build GRCh38
rs924938349 Chr.16: 68061241-68061241 on Build GRCh38
rs543489852 Chr.16: 68061181-68061181 on Build GRCh38
rs529084625 Chr.16: 68061240-68061240 on Build GRCh38
rs922024619 Chr.16: 68061242-68061242 on Build GRCh38
rs969833211 Chr.16: 68061402-68061402 on Build GRCh38
rs764078428 Chr.16: 68061022-68061022 on Build GRCh38
rs770093622 Chr.16: 68061157-68061157 on Build GRCh38
rs551380993 Chr.16: 68061394-68061394 on Build GRCh38
rs985784261 Chr.16: 68061223-68061223 on Build GRCh38
rs769716074 Chr.16: 68061194-68061202 on Build GRCh38
rs376408660 Chr.16: 68061130-68061130 on Build GRCh38
rs747606206 Chr.16: 68061102-68061102 on Build GRCh38
rs112966237 Chr.16: 68061377-68061377 on Build GRCh38
rs1030101903 Chr.16: 68061442-68061442 on Build GRCh38
rs995641874 Chr.16: 68061097-68061097 on Build GRCh38
rs953067890 Chr.16: 68061180-68061180 on Build GRCh38
rs932506285 Chr.16: 68061273-68061273 on Build GRCh38
rs926840666 Chr.16: 68061309-68061309 on Build GRCh38
rs750414038 Chr.16: 68061074-68061074 on Build GRCh38
rs369910078 Chr.16: 68061179-68061179 on Build GRCh38
rs956305466 Chr.16: 68061432-68061432 on Build GRCh38
rs778546604 Chr.16: 68061100-68061100 on Build GRCh38
rs891040482 Chr.16: 68061030-68061030 on Build GRCh38
rs962498498 Chr.16: 68061063-68061063 on Build GRCh38
rs776987845 Chr.16: 68061131-68061131 on Build GRCh38
rs747723178 Chr.16: 68061007-68061007 on Build GRCh38
rs540969294 Chr.16: 68061342-68061342 on Build GRCh38
rs949552703 Chr.16: 68061295-68061295 on Build GRCh38
rs118156865 Chr.16: 68061388-68061388 on Build GRCh38
rs759738135 Chr.16: 68061409-68061409 on Build GRCh38
rs763150190 Chr.16: 68061051-68061051 on Build GRCh38
rs997734051 Chr.16: 68061411-68061411 on Build GRCh38
rs1035787773 Chr.16: 68061422-68061422 on Build GRCh38
rs988295413 Chr.16: 68061290-68061290 on Build GRCh38
rs1027087902 Chr.16: 68061061-68061061 on Build GRCh38
rs200274192 Chr.16: 68061117-68061117 on Build GRCh38
rs1007422536 Chr.16: 68061449-68061449 on Build GRCh38
rs535366136 Chr.16: 68061045-68061045 on Build GRCh38
rs770443853 Chr.16: 68061296-68061296 on Build GRCh38
rs779956332 Chr.16: 68061083-68061083 on Build GRCh38

View Full Product Details