Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 51029844-51030342 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMAD4
Entrez Gene ID
4089
Gene Name
SMAD family member 4
Gene Aliases
DPC4, JIP, MADH4, MYHRS
UniGene
Hs.75862
Gene Chromosome Location
Chr.18: 51030213-51085042 on Build GRCh38
Transcripts
NM_005359.5

SNP Detail

SNP ID Chromosome Location
rs1023021024 Chr.18: 51030170-51030170 on Build GRCh38
rs552784408 Chr.18: 51030298-51030298 on Build GRCh38
rs891331971 Chr.18: 51030275-51030275 on Build GRCh38
rs535203415 Chr.18: 51030308-51030308 on Build GRCh38
rs572855606 Chr.18: 51030301-51030301 on Build GRCh38
rs954415027 Chr.18: 51030057-51030057 on Build GRCh38
rs540072311 Chr.18: 51030281-51030281 on Build GRCh38
rs955474759 Chr.18: 51030113-51030113 on Build GRCh38
rs1051229068 Chr.18: 51030195-51030195 on Build GRCh38
rs945228428 Chr.18: 51030101-51030101 on Build GRCh38
rs1017647643 Chr.18: 51030070-51030070 on Build GRCh38
rs987141251 Chr.18: 51030060-51030060 on Build GRCh38
rs1044087447 Chr.18: 51030083-51030083 on Build GRCh38
rs758154933 Chr.18: 51030097-51030097 on Build GRCh38
rs1017343979 Chr.18: 51030205-51030205 on Build GRCh38
rs939405461 Chr.18: 51030126-51030126 on Build GRCh38
rs928433474 Chr.18: 51030059-51030059 on Build GRCh38
rs891203020 Chr.18: 51030267-51030267 on Build GRCh38
rs960525967 Chr.18: 51030243-51030245 on Build GRCh38
rs906610422 Chr.18: 51029918-51029918 on Build GRCh38
rs994349993 Chr.18: 51030178-51030178 on Build GRCh38
rs915598872 Chr.18: 51030046-51030046 on Build GRCh38
rs900814463 Chr.18: 51030314-51030314 on Build GRCh38
rs955526658 Chr.18: 51029989-51029989 on Build GRCh38
rs1029800440 Chr.18: 51029987-51029987 on Build GRCh38
rs1029748112 Chr.18: 51030111-51030111 on Build GRCh38
rs915095565 Chr.18: 51029890-51029890 on Build GRCh38
rs934494275 Chr.18: 51030069-51030069 on Build GRCh38
rs890211408 Chr.18: 51030200-51030200 on Build GRCh38
rs546363148 Chr.18: 51030304-51030304 on Build GRCh38
rs981114805 Chr.18: 51030207-51030207 on Build GRCh38
rs974366045 Chr.18: 51030163-51030163 on Build GRCh38
rs772538273 Chr.18: 51030292-51030292 on Build GRCh38
rs996952685 Chr.18: 51029986-51029986 on Build GRCh38
rs1038343196 Chr.18: 51030256-51030257 on Build GRCh38
rs145170795 Chr.18: 51029892-51029892 on Build GRCh38
rs1038952076 Chr.18: 51030237-51030237 on Build GRCh38
rs1022653162 Chr.18: 51030221-51030221 on Build GRCh38
rs193246988 Chr.18: 51030106-51030106 on Build GRCh38
rs867046120 Chr.18: 51030236-51030236 on Build GRCh38
rs1051342921 Chr.18: 51030271-51030271 on Build GRCh38
rs886053885 Chr.18: 51030248-51030250 on Build GRCh38
rs1015794051 Chr.18: 51030133-51030133 on Build GRCh38
rs897380467 Chr.18: 51030165-51030165 on Build GRCh38
rs528465246 Chr.18: 51030088-51030088 on Build GRCh38
rs552365456 Chr.18: 51030040-51030040 on Build GRCh38
rs948446878 Chr.18: 51030047-51030047 on Build GRCh38
rs934442083 Chr.18: 51030211-51030211 on Build GRCh38
rs754733653 Chr.18: 51030008-51030008 on Build GRCh38
rs1045465134 Chr.18: 51030052-51030052 on Build GRCh38
rs886053887 Chr.18: 51030275-51030275 on Build GRCh38
rs908785899 Chr.18: 51030026-51030026 on Build GRCh38
rs900453280 Chr.18: 51030247-51030247 on Build GRCh38
rs1039048347 Chr.18: 51030162-51030162 on Build GRCh38
rs1051119571 Chr.18: 51030256-51030256 on Build GRCh38
rs570695602 Chr.18: 51030268-51030268 on Build GRCh38
rs914422902 Chr.18: 51030233-51030233 on Build GRCh38
rs1004371406 Chr.18: 51030125-51030125 on Build GRCh38
rs1055021921 Chr.18: 51029972-51029972 on Build GRCh38
rs941358039 Chr.18: 51030158-51030158 on Build GRCh38
rs763537410 Chr.18: 51030321-51030321 on Build GRCh38
rs753783580 Chr.18: 51029917-51029917 on Build GRCh38
rs988578345 Chr.18: 51030220-51030220 on Build GRCh38
rs990898741 Chr.18: 51030246-51030246 on Build GRCh38
rs540145914 Chr.18: 51030015-51030016 on Build GRCh38
rs932637142 Chr.18: 51030252-51030252 on Build GRCh38
rs963021931 Chr.18: 51030038-51030038 on Build GRCh38
rs1055050915 Chr.18: 51030135-51030135 on Build GRCh38
rs886053886 Chr.18: 51030256-51030256 on Build GRCh38
rs939374562 Chr.18: 51029931-51029931 on Build GRCh38
rs189177661 Chr.18: 51030071-51030071 on Build GRCh38
rs112585023 Chr.18: 51030077-51030077 on Build GRCh38
rs911207966 Chr.18: 51030322-51030344 on Build GRCh38
rs945224414 Chr.18: 51029916-51029916 on Build GRCh38
rs898665509 Chr.18: 51030312-51030312 on Build GRCh38
rs969754589 Chr.18: 51030206-51030206 on Build GRCh38
rs895408281 Chr.18: 51029977-51029977 on Build GRCh38
rs557246609 Chr.18: 51030251-51030251 on Build GRCh38
rs537319496 Chr.18: 51030219-51030219 on Build GRCh38
rs147613425 Chr.18: 51030140-51030140 on Build GRCh38
rs1008688273 Chr.18: 51030203-51030203 on Build GRCh38
rs778581748 Chr.18: 51030044-51030044 on Build GRCh38
rs994318437 Chr.18: 51030045-51030045 on Build GRCh38
rs752342578 Chr.18: 51030075-51030075 on Build GRCh38
rs899838010 Chr.18: 51030103-51030103 on Build GRCh38
rs777608613 Chr.18: 51030208-51030208 on Build GRCh38

View Full Product Details