Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 81705266-81705805 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HGF
Entrez Gene ID
3082
Gene Name
hepatocyte growth factor
Gene Aliases
DFNB39, F-TCF, HGFB, HPTA, SF
UniGene
Hs.396530
Gene Chromosome Location
Chr.7: 81699006-81770438 on Build GRCh38
Transcripts
NM_000601.5 NM_001010932.2 XM_006715956.2 XM_011516115.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1488699 c.1913G>A p.G638E Chr.7: 81705487-81705487 on Build GRCh38
6515099 c.1876G>T p.D626Y Chr.7: 81705524-81705524 on Build GRCh38
6908827 c.1926C>A p.C642* Chr.7: 81705474-81705474 on Build GRCh38
7371068 c.1813A>T p.T605S Chr.7: 81705698-81705698 on Build GRCh38
6679664 c.1889G>A p.R630Q Chr.7: 81705511-81705511 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778863051 Chr.7: 81705757-81705757 on Build GRCh38
rs376395424 Chr.7: 81705534-81705534 on Build GRCh38
rs747052903 Chr.7: 81705604-81705604 on Build GRCh38
rs747158538 Chr.7: 81705511-81705511 on Build GRCh38
rs764695573 Chr.7: 81705552-81705552 on Build GRCh38
rs780359807 Chr.7: 81705602-81705602 on Build GRCh38
rs746110664 Chr.7: 81705502-81705502 on Build GRCh38
rs200374420 Chr.7: 81705654-81705654 on Build GRCh38
rs771961342 Chr.7: 81705775-81705775 on Build GRCh38
rs540676769 Chr.7: 81705542-81705542 on Build GRCh38
rs769580628 Chr.7: 81705419-81705419 on Build GRCh38
rs757640030 Chr.7: 81705560-81705560 on Build GRCh38
rs369418750 Chr.7: 81705456-81705456 on Build GRCh38
rs769920904 Chr.7: 81705579-81705582 on Build GRCh38
rs954312187 Chr.7: 81705522-81705522 on Build GRCh38
rs371548104 Chr.7: 81705344-81705344 on Build GRCh38
rs765890500 Chr.7: 81705695-81705695 on Build GRCh38
rs375394415 Chr.7: 81705351-81705351 on Build GRCh38
rs998501328 Chr.7: 81705318-81705321 on Build GRCh38
rs145494248 Chr.7: 81705509-81705509 on Build GRCh38
rs770672389 Chr.7: 81705649-81705649 on Build GRCh38
rs761664295 Chr.7: 81705410-81705410 on Build GRCh38
rs750636675 Chr.7: 81705566-81705566 on Build GRCh38
rs778031781 Chr.7: 81705378-81705378 on Build GRCh38
rs141774517 Chr.7: 81705679-81705679 on Build GRCh38
rs184717721 Chr.7: 81705561-81705561 on Build GRCh38
rs199516810 Chr.7: 81705738-81705738 on Build GRCh38
rs772967560 Chr.7: 81705421-81705421 on Build GRCh38
rs755575139 Chr.7: 81705531-81705531 on Build GRCh38
rs150109563 Chr.7: 81705458-81705458 on Build GRCh38
rs926889688 Chr.7: 81705545-81705545 on Build GRCh38
rs878880281 Chr.7: 81705597-81705597 on Build GRCh38
rs775281878 Chr.7: 81705663-81705663 on Build GRCh38
rs781145595 Chr.7: 81705342-81705342 on Build GRCh38
rs777216192 Chr.7: 81705682-81705682 on Build GRCh38
rs756602776 Chr.7: 81705481-81705481 on Build GRCh38
rs373545882 Chr.7: 81705528-81705528 on Build GRCh38
rs772105726 Chr.7: 81705393-81705393 on Build GRCh38
rs766007676 Chr.7: 81705435-81705435 on Build GRCh38
rs748539273 Chr.7: 81705593-81705593 on Build GRCh38
rs545249104 Chr.7: 81705461-81705461 on Build GRCh38
rs565056780 Chr.7: 81705496-81705496 on Build GRCh38
rs769585774 Chr.7: 81705512-81705512 on Build GRCh38
rs202215700 Chr.7: 81705466-81705466 on Build GRCh38
rs751819844 Chr.7: 81705724-81705724 on Build GRCh38
rs766493585 Chr.7: 81705723-81705723 on Build GRCh38
rs779827905 Chr.7: 81705771-81705776 on Build GRCh38
rs779267373 Chr.7: 81705500-81705500 on Build GRCh38
rs931329793 Chr.7: 81705622-81705622 on Build GRCh38
rs147075806 Chr.7: 81705697-81705697 on Build GRCh38
rs745942461 Chr.7: 81705384-81705384 on Build GRCh38
rs553489977 Chr.7: 81705354-81705354 on Build GRCh38
rs749474633 Chr.7: 81705380-81705380 on Build GRCh38
rs777627774 Chr.7: 81705635-81705635 on Build GRCh38
rs779318616 Chr.7: 81705382-81705382 on Build GRCh38
rs762445545 Chr.7: 81705693-81705693 on Build GRCh38
rs536246454 Chr.7: 81705366-81705366 on Build GRCh38
rs778395987 Chr.7: 81705483-81705483 on Build GRCh38
rs758523692 Chr.7: 81705582-81705582 on Build GRCh38
rs751270979 Chr.7: 81705446-81705446 on Build GRCh38
rs774153258 Chr.7: 81705652-81705652 on Build GRCh38
rs758862083 Chr.7: 81705718-81705718 on Build GRCh38
rs754944443 Chr.7: 81705735-81705735 on Build GRCh38
rs370292193 Chr.7: 81705768-81705768 on Build GRCh38
rs1028646568 Chr.7: 81705734-81705734 on Build GRCh38
rs745648975 Chr.7: 81705761-81705761 on Build GRCh38
rs13235690 Chr.7: 81705626-81705626 on Build GRCh38
rs770723689 Chr.7: 81705520-81705520 on Build GRCh38
rs745642167 Chr.7: 81705382-81705383 on Build GRCh38
rs754226152 Chr.7: 81705494-81705494 on Build GRCh38
rs781715040 Chr.7: 81705361-81705361 on Build GRCh38
rs771594786 Chr.7: 81705658-81705658 on Build GRCh38
rs776576073 Chr.7: 81705406-81705406 on Build GRCh38
rs754457294 Chr.7: 81705557-81705557 on Build GRCh38
rs749226169 Chr.7: 81705637-81705637 on Build GRCh38
rs773198515 Chr.7: 81705515-81705515 on Build GRCh38
rs750048589 Chr.7: 81705350-81705350 on Build GRCh38
rs900830869 Chr.7: 81705719-81705719 on Build GRCh38
rs531850775 Chr.7: 81705747-81705747 on Build GRCh38
rs866398503 Chr.7: 81705645-81705645 on Build GRCh38
rs755141326 Chr.7: 81705619-81705619 on Build GRCh38
rs371826587 Chr.7: 81705533-81705533 on Build GRCh38
rs201730498 Chr.7: 81705381-81705381 on Build GRCh38
rs1012056012 Chr.7: 81705448-81705448 on Build GRCh38
rs192932545 Chr.7: 81705369-81705369 on Build GRCh38
rs760093179 Chr.7: 81705673-81705673 on Build GRCh38
rs780620898 Chr.7: 81705333-81705333 on Build GRCh38
rs13238580 Chr.7: 81705627-81705627 on Build GRCh38
rs187442152 Chr.7: 81705335-81705335 on Build GRCh38
rs766064717 Chr.7: 81705353-81705353 on Build GRCh38
rs5745755 Chr.7: 81705332-81705332 on Build GRCh38
rs538415452 Chr.7: 81705746-81705746 on Build GRCh38
rs200169107 Chr.7: 81705359-81705359 on Build GRCh38
rs762721825 Chr.7: 81705516-81705516 on Build GRCh38
rs779495509 Chr.7: 81705547-81705547 on Build GRCh38

View Full Product Details