Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 144511564-144512066 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RECQL4
Entrez Gene ID
9401
Gene Name
RecQ like helicase 4
Gene Aliases
RECQ4
UniGene
Hs.31442
Gene Chromosome Location
Chr.8: 144511284-144517833 on Build GRCh38
Transcripts
NM_004260.3 XM_011517384.2 XM_017013991.1 XM_017013992.1 XM_017013993.1 XM_017013994.1 XM_017013995.1 XM_017013996.1 XM_017013997.1 XM_017013998.1 XM_017013999.1 XM_017014000.1 XM_017014001.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6982726 c.3497G>T p.G1166V Chr.8: 144511686-144511686 on Build GRCh38
6951617 c.3424G>A p.D1142N Chr.8: 144511759-144511759 on Build GRCh38
4762358 c.3286C>T p.R1096C Chr.8: 144512018-144512018 on Build GRCh38
3685354 c.3394C>G p.L1132V Chr.8: 144511789-144511789 on Build GRCh38
4762359 c.3281A>C p.E1094A Chr.8: 144512023-144512023 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs878854649 Chr.8: 144511902-144511903 on Build GRCh38
rs749208991 Chr.8: 144511929-144511929 on Build GRCh38
rs779196557 Chr.8: 144512025-144512026 on Build GRCh38
rs768802673 Chr.8: 144512045-144512045 on Build GRCh38
rs916377054 Chr.8: 144511861-144511861 on Build GRCh38
rs747815189 Chr.8: 144511698-144511698 on Build GRCh38
rs575014753 Chr.8: 144511612-144511612 on Build GRCh38
rs2279243 Chr.8: 144511657-144511657 on Build GRCh38
rs1056877904 Chr.8: 144511880-144511880 on Build GRCh38
rs757563486 Chr.8: 144511680-144511680 on Build GRCh38
rs373689479 Chr.8: 144511882-144511882 on Build GRCh38
rs773909942 Chr.8: 144511952-144511952 on Build GRCh38
rs767822615 Chr.8: 144511764-144511764 on Build GRCh38
rs766056345 Chr.8: 144512009-144512009 on Build GRCh38
rs535054133 Chr.8: 144511871-144511871 on Build GRCh38
rs774386060 Chr.8: 144511805-144511805 on Build GRCh38
rs4251692 Chr.8: 144511902-144511902 on Build GRCh38
rs587778646 Chr.8: 144512048-144512048 on Build GRCh38
rs1056576166 Chr.8: 144511694-144511694 on Build GRCh38
rs779410033 Chr.8: 144511687-144511687 on Build GRCh38
rs775431574 Chr.8: 144511710-144511710 on Build GRCh38
rs770697720 Chr.8: 144511799-144511799 on Build GRCh38
rs765087683 Chr.8: 144512017-144512017 on Build GRCh38
rs371049072 Chr.8: 144512018-144512018 on Build GRCh38
rs781456397 Chr.8: 144511986-144511986 on Build GRCh38
rs779826649 Chr.8: 144511928-144511928 on Build GRCh38
rs557284122 Chr.8: 144511909-144511909 on Build GRCh38
rs751187872 Chr.8: 144511835-144511835 on Build GRCh38
rs751838450 Chr.8: 144511826-144511826 on Build GRCh38
rs35412242 Chr.8: 144511639-144511639 on Build GRCh38
rs376392965 Chr.8: 144511983-144511983 on Build GRCh38
rs990831417 Chr.8: 144511962-144511962 on Build GRCh38
rs75312128 Chr.8: 144511812-144511812 on Build GRCh38
rs766585337 Chr.8: 144511815-144511815 on Build GRCh38
rs377390897 Chr.8: 144511943-144511943 on Build GRCh38
rs774202918 Chr.8: 144511690-144511690 on Build GRCh38
rs995015088 Chr.8: 144511937-144511937 on Build GRCh38
rs759478327 Chr.8: 144511755-144511755 on Build GRCh38
rs773195667 Chr.8: 144511878-144511878 on Build GRCh38
rs754479731 Chr.8: 144511739-144511739 on Build GRCh38
rs751080188 Chr.8: 144511904-144511904 on Build GRCh38
rs572936673 Chr.8: 144511966-144511966 on Build GRCh38
rs760094869 Chr.8: 144511824-144511824 on Build GRCh38
rs779060469 Chr.8: 144511603-144511603 on Build GRCh38
rs922998248 Chr.8: 144511646-144511646 on Build GRCh38
rs758521652 Chr.8: 144511676-144511676 on Build GRCh38
rs767603116 Chr.8: 144511590-144511590 on Build GRCh38
rs750267463 Chr.8: 144511662-144511662 on Build GRCh38
rs879073921 Chr.8: 144511642-144511642 on Build GRCh38
rs758435669 Chr.8: 144512026-144512026 on Build GRCh38
rs753620447 Chr.8: 144511970-144511970 on Build GRCh38
rs752301125 Chr.8: 144511790-144511790 on Build GRCh38
rs754909617 Chr.8: 144511974-144511974 on Build GRCh38
rs950715422 Chr.8: 144511773-144511773 on Build GRCh38
rs766724974 Chr.8: 144511819-144511819 on Build GRCh38
rs35995510 Chr.8: 144511827-144511827 on Build GRCh38
rs762384825 Chr.8: 144511999-144511999 on Build GRCh38
rs202083938 Chr.8: 144511688-144511688 on Build GRCh38
rs752841311 Chr.8: 144511665-144511665 on Build GRCh38
rs35346077 Chr.8: 144511740-144511740 on Build GRCh38
rs371037028 Chr.8: 144511781-144511781 on Build GRCh38
rs751332709 Chr.8: 144511680-144511681 on Build GRCh38
rs757342740 Chr.8: 144511985-144511985 on Build GRCh38
rs916484589 Chr.8: 144511702-144511702 on Build GRCh38
rs868706147 Chr.8: 144512007-144512007 on Build GRCh38
rs773088003 Chr.8: 144511699-144511699 on Build GRCh38
rs781403453 Chr.8: 144512027-144512027 on Build GRCh38
rs756882629 Chr.8: 144511864-144511864 on Build GRCh38
rs370611564 Chr.8: 144511760-144511760 on Build GRCh38
rs575840682 Chr.8: 144511664-144511664 on Build GRCh38
rs61755066 Chr.8: 144511748-144511748 on Build GRCh38
rs747417851 Chr.8: 144511632-144511632 on Build GRCh38
rs781543298 Chr.8: 144511681-144511681 on Build GRCh38
rs755845445 Chr.8: 144511921-144511921 on Build GRCh38
rs756549605 Chr.8: 144511727-144511727 on Build GRCh38
rs368712314 Chr.8: 144511956-144511956 on Build GRCh38
rs137853230 Chr.8: 144512033-144512033 on Build GRCh38
rs34915097 Chr.8: 144511991-144511991 on Build GRCh38
rs372943506 Chr.8: 144511660-144511660 on Build GRCh38
rs775893146 Chr.8: 144511989-144511989 on Build GRCh38
rs756627 Chr.8: 144511903-144511903 on Build GRCh38
rs1042489227 Chr.8: 144511901-144511901 on Build GRCh38
rs757224601 Chr.8: 144511686-144511686 on Build GRCh38
rs766385605 Chr.8: 144511707-144511707 on Build GRCh38
rs34375336 Chr.8: 144511827-144511828 on Build GRCh38
rs562254930 Chr.8: 144511976-144511976 on Build GRCh38
rs868635592 Chr.8: 144511964-144511964 on Build GRCh38
rs754952505 Chr.8: 144511878-144511878 on Build GRCh38
rs367836099 Chr.8: 144511885-144511885 on Build GRCh38
rs771428532 Chr.8: 144511633-144511633 on Build GRCh38
rs754328779 Chr.8: 144511772-144511772 on Build GRCh38
rs202134651 Chr.8: 144511753-144511753 on Build GRCh38
rs770246912 Chr.8: 144511944-144511944 on Build GRCh38
rs780986647 Chr.8: 144511724-144511726 on Build GRCh38
rs769772821 Chr.8: 144511803-144511803 on Build GRCh38
rs770193726 Chr.8: 144511988-144511988 on Build GRCh38
rs973395295 Chr.8: 144511645-144511645 on Build GRCh38
rs747758347 Chr.8: 144511747-144511747 on Build GRCh38
rs201369291 Chr.8: 144511912-144511912 on Build GRCh38
rs910228017 Chr.8: 144511611-144511611 on Build GRCh38
rs992381749 Chr.8: 144511977-144511977 on Build GRCh38
rs35626651 Chr.8: 144511687-144511688 on Build GRCh38
rs750008283 Chr.8: 144511959-144511959 on Build GRCh38
rs776076152 Chr.8: 144511894-144511894 on Build GRCh38
rs760298099 Chr.8: 144512008-144512008 on Build GRCh38
rs533732397 Chr.8: 144511866-144511866 on Build GRCh38
rs986482653 Chr.8: 144511995-144511995 on Build GRCh38
rs35101495 Chr.8: 144511967-144511967 on Build GRCh38
rs756666498 Chr.8: 144511685-144511685 on Build GRCh38
rs768472726 Chr.8: 144511649-144511649 on Build GRCh38
rs748732922 Chr.8: 144511738-144511738 on Build GRCh38
rs148752485 Chr.8: 144511941-144511941 on Build GRCh38
rs761949478 Chr.8: 144511806-144511806 on Build GRCh38
rs368573846 Chr.8: 144511867-144511867 on Build GRCh38
rs772122711 Chr.8: 144511874-144511874 on Build GRCh38
rs376559433 Chr.8: 144511920-144511920 on Build GRCh38
rs780200030 Chr.8: 144512041-144512041 on Build GRCh38
rs36078464 Chr.8: 144511990-144511990 on Build GRCh38
rs763496147 Chr.8: 144511947-144511947 on Build GRCh38
rs375432640 Chr.8: 144511765-144511765 on Build GRCh38
rs767821367 Chr.8: 144511722-144511722 on Build GRCh38
rs369843844 Chr.8: 144511940-144511940 on Build GRCh38
rs777156550 Chr.8: 144511829-144511829 on Build GRCh38
rs751909716 Chr.8: 144511678-144511678 on Build GRCh38
rs370677156 Chr.8: 144511800-144511800 on Build GRCh38
rs1043628799 Chr.8: 144511708-144511708 on Build GRCh38
rs372315574 Chr.8: 144511881-144511881 on Build GRCh38
rs746403783 Chr.8: 144511634-144511634 on Build GRCh38
rs746656592 Chr.8: 144511798-144511798 on Build GRCh38
rs776023102 Chr.8: 144511946-144511946 on Build GRCh38
rs777101999 Chr.8: 144511884-144511884 on Build GRCh38
rs371296496 Chr.8: 144511604-144511604 on Build GRCh38
rs755069201 Chr.8: 144511598-144511598 on Build GRCh38
rs587778647 Chr.8: 144511762-144511762 on Build GRCh38
rs777884007 Chr.8: 144511932-144511932 on Build GRCh38
rs529113837 Chr.8: 144511754-144511754 on Build GRCh38
rs539751764 Chr.8: 144511802-144511802 on Build GRCh38
rs780676589 Chr.8: 144511734-144511734 on Build GRCh38
rs898264400 Chr.8: 144512021-144512021 on Build GRCh38
rs778513153 Chr.8: 144511784-144511784 on Build GRCh38
rs34914400 Chr.8: 144511703-144511703 on Build GRCh38
rs767643917 Chr.8: 144511818-144511818 on Build GRCh38
rs746323102 Chr.8: 144511682-144511682 on Build GRCh38
rs562024579 Chr.8: 144511751-144511751 on Build GRCh38
rs757502009 Chr.8: 144511720-144511720 on Build GRCh38
rs1030415007 Chr.8: 144511927-144511927 on Build GRCh38
rs774256449 Chr.8: 144511650-144511650 on Build GRCh38
rs764549432 Chr.8: 144511948-144511948 on Build GRCh38
rs759304160 Chr.8: 144512015-144512015 on Build GRCh38
rs950777473 Chr.8: 144511658-144511658 on Build GRCh38
rs758089563 Chr.8: 144511791-144511791 on Build GRCh38
rs749540920 Chr.8: 144512043-144512043 on Build GRCh38
rs756401653 Chr.8: 144512036-144512036 on Build GRCh38
rs763080171 Chr.8: 144511715-144511715 on Build GRCh38
rs903928753 Chr.8: 144511992-144511992 on Build GRCh38
rs752540498 Chr.8: 144512022-144512022 on Build GRCh38
rs34236392 Chr.8: 144511987-144511987 on Build GRCh38
rs188078422 Chr.8: 144511828-144511828 on Build GRCh38
rs754273420 Chr.8: 144511821-144511821 on Build GRCh38
rs376874393 Chr.8: 144511718-144511718 on Build GRCh38
rs777542762 Chr.8: 144511793-144511793 on Build GRCh38
rs371623716 Chr.8: 144511891-144511891 on Build GRCh38
rs753659106 Chr.8: 144512010-144512010 on Build GRCh38
rs61755067 Chr.8: 144511759-144511759 on Build GRCh38
rs908693736 Chr.8: 144511671-144511671 on Build GRCh38
rs564143545 Chr.8: 144511726-144511726 on Build GRCh38
rs746892308 Chr.8: 144511752-144511752 on Build GRCh38
rs769119630 Chr.8: 144511637-144511637 on Build GRCh38
rs771528925 Chr.8: 144512001-144512001 on Build GRCh38
rs758674099 Chr.8: 144511605-144511605 on Build GRCh38
rs761818423 Chr.8: 144511587-144511587 on Build GRCh38
rs761157177 Chr.8: 144511766-144511766 on Build GRCh38
rs368671582 Chr.8: 144511957-144511957 on Build GRCh38
rs772729762 Chr.8: 144512002-144512002 on Build GRCh38
rs930778534 Chr.8: 144512030-144512030 on Build GRCh38
rs753960282 Chr.8: 144511593-144511593 on Build GRCh38
rs762463899 Chr.8: 144511640-144511640 on Build GRCh38
rs1017854921 Chr.8: 144511618-144511618 on Build GRCh38
rs373652049 Chr.8: 144511716-144511716 on Build GRCh38

View Full Product Details